Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
The Pierce BCA Protein Assay Kit is a high-precision, detergent-compatible protein assay for determination of protein concentration. Pierce BCA reagents provide accurate determination of protein concentration with most sample types encountered in protein research. The Pierce BCA assay can be used to assess yields in whole cell lysates, affinity-column fractions, purified proteins samples, as well as to monitor protein contamination in industrial applications. Compared to most dye-binding methods, the BCA assay is affected much less by protein compositional differences, providing greater concentration accuracy.
Kits are available with or without Pierce Dilution-Free BSA Protein Standards, which are a set of seven pre-diluted BSA standards, packaged in a multichannel pipette-friendly tubestrip. The tubestrip includes a single empty tube that enables users to add their own sample buffer for the purpose of blank subtraction.
Concentration | BSA: 0.125, 0.25, 0.5, 0.75, 1, 1.5, and 2 mg/mL |
Description | Pierce BCA Protein Assay Kit with Dilution-Free BSA Protein Standards |
Assay | BCA Assay |
Detection Method | Colorimetric |
Product Line | Pierce™ |
Product Type | Protein Quantitation Assay |
Specificity | Not Target-specific |
Sufficient For | 500 Tube Assays or 5000 Microplate Assays |
For Use With (Application) | Solution-based Detection, Absorbance |
For Use With (Equipment) | Spectrophotometer, Microplate Reader |
Kit Contents | 4 x 1 mL 8-channel tubestrip, prediluted |
Quantity | 1 Kit (1 L) |
Unit Size | 1 L |
Concentration | BSA: 2 mg/mL |
Description | Pierce BCA Protein Assay Kit, 100 mL |
Assay | BCA Assay |
Detection Method | Colorimetric |
Product Line | Pierce™ |
Product Type | Protein Quantitation Assay |
Specificity | Not Target-Specific |
Sufficient For | 50 Tube Assays or 500 Microplate Assays |
For Use With (Application) | Solution-based Detection, Absorbance |
For Use With (Equipment) | Spectrophotometer, Microplate Reader |
Kit Contents | 2 mL vial |
Quantity | 1 Kit (100 mL) |
Unit Size | 100 mL |
Concentration | BSA: 2 mg/mL |
Description | Pierce BCA Protein Assay Kit, 1000 mL |
Assay | BCA Assay |
Detection Method | Colorimetric |
Product Line | Pierce™ |
Product Type | Protein Quantitation Assay |
Specificity | Not Target-Specific |
Sufficient For | 500 Tube Assays or 5000 Microplate Assays |
For Use With (Application) | Solution-based Detection, Absorbance |
For Use With (Equipment) | Spectrophotometer, Microplate Reader |
Kit Contents | 10 x 1 mL glass ampules |
Quantity | 1 Kit (1 L) |
Unit Size | 1 L |
Concentration | BSA: 2 mg/mL |
Description | Pierce BCA Protein Assay Kit, 500 mL |
Assay | BCA Assay |
Detection Method | Colorimetric |
Product Line | Pierce™ |
Product Type | Protein Quantitation Assay |
Specificity | Not Target-Specific |
Sufficient For | 250 Tube Assays or 2500 Microplate Assays |
For Use With (Application) | Solution-based Detection, Absorbance |
For Use With (Equipment) | Spectrophotometer, Microplate Reader |
Kit Contents | 10 x 1 mL glass ampules |
Quantity | 1 Kit (500 mL) |
Unit Size | 500 mL |
Concentration | BSA: 0.125, 0.25, 0.5, 0.75, 1, 1.5, and 2 mg/mL |
Description | Pierce BCA Protein Assay Kit with Dilution-Free BSA Protein Standards |
Assay | BCA Assay |
Detection Method | Colorimetric |
Product Line | Pierce™ |
Product Type | Protein Quantitation Assay |
Specificity | Not Target-specific |
Sufficient For | 250 Tube Assays or 2500 Microplate Assays |
For Use With (Application) | Solution-based Detection, Absorbance |
For Use With (Equipment) | Spectrophotometer, Microplate Reader |
Kit Contents | 2 x 1 mL 8-channel tubestrip, prediluted |
Quantity | 1 Kit (500 mL) |
Unit Size | 500 mL |
Catalog Number | Specifications | Unit Size | Description | Concentration | Kit Contents | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
A55864 | 1 L | Pierce BCA Protein Assay Kit with Dilution-Free BSA Protein Standards | BSA: 0.125, 0.25, 0.5, 0.75, 1, 1.5, and 2 mg/mL | 4 x 1 mL 8-channel tubestrip, prediluted | 305.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A65453 | 100 mL | Pierce BCA Protein Assay Kit, 100 mL | BSA: 2 mg/mL | 2 mL vial | 66.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
23225 | 1 L | Pierce BCA Protein Assay Kit, 1000 mL | BSA: 2 mg/mL | 10 x 1 mL glass ampules | 234.65 Online price 254.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
23227 | 500 mL | Pierce BCA Protein Assay Kit, 500 mL | BSA: 2 mg/mL | 10 x 1 mL glass ampules | 151.65 Online price 163.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
A55865 | 500 mL | Pierce BCA Protein Assay Kit with Dilution-Free BSA Protein Standards | BSA: 0.125, 0.25, 0.5, 0.75, 1, 1.5, and 2 mg/mL | 2 x 1 mL 8-channel tubestrip, prediluted | 215.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
The Pierce BCA Protein Assay Kit is a high-precision, detergent-compatible protein assay for determination of protein concentration. Pierce BCA reagents provide accurate determination of protein concentration with most sample types encountered in protein research. The Pierce BCA assay can be used to assess yields in whole cell lysates, affinity-column fractions, purified proteins samples, as well as to monitor protein contamination in industrial applications. Compared to most dye-binding methods, the BCA assay is affected much less by protein compositional differences, providing greater concentration accuracy.
Kit options are available that include Pierce Dilution-Free BSA Protein Standards, which are a set of seven pre-diluted BSA standards, packaged in a multichannel pipette-friendly tubestrip. The tubestrip includes a single empty tube that enables users to add their own sample buffer for the purpose of blank subtraction.
Features of the BCA Protein Assay Kit include:
How the assay works
The BCA Protein Assay combines the well-known reduction of Cu2+ to Cu1+ by protein in an alkaline medium with the highly sensitive and selective colorimetric detection of the cuprous cation (Cu1+) by bicinchoninic acid (BCA). The first step is the chelation of copper with protein in an alkaline environment to form a light blue complex. In this reaction, known as the biuret reaction, peptides containing three or more amino acid residues form a colored chelate complex with cupric ions in an alkaline environment containing sodium potassium tartrate.
In the second step of the color development reaction, BCA reacts with the reduced (cuprous) cation that was formed in step one. The intense purple-colored reaction product results from the chelation of two molecules of BCA with one cuprous ion. The BCA/copper complex is water soluble and exhibits a strong linear absorbance at 562 nm with increasing protein concentrations. The complex is approximately 100 times more sensitive (lower limit of detection) than the pale blue color of the first reaction.
The reaction is strongly influenced by four amino acid residues (cysteine, cystine, tyrosine, and tryptophan) in the amino acid sequence of the protein. However, unlike Coomassie dye-binding methods (i.e., Bradford), the universal peptide backbone also contributes to color formation, helping to minimize variability caused by protein compositional differences.