Informational:Informational:Save up to 31% on your order of cell culture products. View promotion details »
Gibco™

Fetal Bovine Serum (FBS), Premium, One Shot™ format

Catalog number: A5670402
Have Questions?
Gibco™

Fetal Bovine Serum (FBS), Premium, One Shot™ format

Catalog number: A5670402
Have Questions?
Catalog Number
A5670402
Unit Size
10 x 50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
50 mL
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
500 mL
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A567040210 x 50 mL
-
A567040150 mL
-
A5670701500 mL
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Gibco sera: trusted quality for consistent performance

Sera category: Premium

  • Use for general cell culture with common cell lines
  • Low endotoxin and hemoglobin levels
  • Endotoxin level: <10 EU/mL
  • Hemoglobin level: <25 mg/dL
  • Origin: refer to CoA for specific origin. All products may not be available in all regions due to importation regulations.
  • US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US

Gibco serum delivers

  • ISO 13485–certified; processed in FDA registered facilities
  • Triple-filtered at 0.1 μm
  • Gibco bottle easier to use in hood; helps reduce risk of contamination and improve consistency
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

Specifications

Age
Fetal
Species
Cattle/Bovine
Country of Origin
Refer to CoA for specific origin
Form
Frozen
Format
One Shot
Product Type
Fetal Bovine Serum
Quantity
10 x 50 mL
Sterility
Sterile-filtered
Sterilization Method
Triple-filtered, 0.1 μm
Grade
Premium
Tested For
Up to 60 quality tests including 9CFR virus testing, endotoxin, and performance
Endotoxin Concentration
≤10 EU/mL
Green Features
Less waste
Hemoglobin Concentration
≤25 mg/dL
Purity or Quality Grade
Premium
Shelf Life
5 Years
Shipping Condition
Dry Ice

Contents & Storage

Store at ≤-10°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text