VetMAX™ Rotavirus A/B/C Controls
VetMAX™ Rotavirus A/B/C Controls
Applied Biosystems™

VetMAX™ Rotavirus A/B/C Controls

VetMAX Rotavirus A/B/C Controls are for use in multiplex, reverse-transcription real-time PCR (RT-qPCR) workflows targeting rotavirus subtypes A, B, andRead more
Have Questions?
Catalog NumberQuantity
A5745110 reactions
Catalog number A57451
Price (USD)
148.00
Each
-
Add to cart
Quantity:
10 reactions
Price (USD)
148.00
Each
Add to cart
Ask our AI about this Product

VetMAX Rotavirus A/B/C Controls are for use in multiplex, reverse-transcription real-time PCR (RT-qPCR) workflows targeting rotavirus subtypes A, B, and C and Xeno RNA control. The controls contain single- and double-stranded RNA and come pre-mixed in one tube as a ready-to-use solution. The included Xeno RNA control (1,000 c/μL) is an internal positive control added to the lysis solution during RNA isolation to distinguish PCR inhibition, reaction failure, or human error from true negatives.

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Concentration1X
DiseasePorcine Rotavirus
No. of Reactions10 reactions
Quantity10 reactions
Shelf Life12 Months
Shipping ConditionShipped at -30°C to -10°C
For Use With (Application)Real Time PCR (qPCR)
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2405002Certificate of AnalysisApr 30, 2025A57451
2502004Certificate of AnalysisMar 20, 2025A57451
2308001Certificate of AnalysisDec 24, 2024A57451
2407003Certificate of AnalysisSep 16, 2024A57451
4 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection