CTS™ OpTmizer™ One SFM
CTS™ OpTmizer™ One SFM
Gibco™

CTS™ OpTmizer™ One SFM

Gibco CTS OpTmizer One SFM is a novel animal-origin-free (AOF) serum-free medium (SFM) formulation developed for the growth and expansion of human T lymphocytes used in cell therapies.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFormatQuantity
A5757203BPC5 L
A5757201Bottle1 L
A5757202BPC1 L
A5757204BPC10 L
4 Options
Catalog number A5757203
Price (USD)/ Each
2,796.00
-
Add to cart
Format:
BPC
Quantity:
5 L
Price (USD)/ Each
2,796.00
Add to cart
CTS™ OpTmizer™ One SFM
Catalog numberA5757203
Price (USD)/ Each
2,796.00
-
Add to cart
Gibco CTS OpTmizer One SFM is a novel animal-origin-free (AOF) serum-free medium (SFM) formulation developed for the growth and expansion of human T lymphocytes used in cell therapies. This medium improves cell count and early memory phenotype throughout all stages of expansion.

Benefits:

  • Provides enhanced T-cell expansion and expression of early memory phenotype makers compared to other chemically defined media
  • Demonstrates flexibility across culture vessels. In both G-Rex™ plates and rocking bioreactors, it supports scale-up while maintaining desired T-cell phenotype.
  • Expands both healthy and disease state donors
  • Supports efficient viral and non-viral editing with downstream expansion
  • Mitigates risk of contaminants and variability associated with animal-sourced materials
  • Supports scale-up via larger catalog pack sizes

Features:

  • Animal-origin-free (AOF) formulation
  • Serum-free—designed without the need for serum or serum replacement
  • No separate components—no need to mix basal medium with supplement to obtain complete medium
  • Yields high density cultures
  • Supports T-cell activation using CTS Dynabeads (both passive release CD3/CD28 (Cat. No.40203D) and detachable CD3/CD28 (Cat No. A56992))
  • Available in 1 L bottle, 1 L bag, 5 L bag, and 10 L bag options

Increased efficiency and consistent performance:

CTS OpTmizer One SFM is an AOF formulation that supports high density cultures in static and dynamic vessels. It displays consistent performance for T-cell expansion, with better early memory phenotype at early stages.

Simplify your transition into clinical research:

The Gibco Cell Therapy Systems (CTS) portfolio offers a broad array of products for use in cell therapy applications, including media, reagents, growth factors, enzymes, cell selection/expansion reagents, and devices. CTS products deliver:

  • Reduced burden in qualifying reagents
  • Appropriate certificates of analysis and origin available
  • Convenient access to our Drug Master File (DMF)
For Research Use or Manufacturing of Cell, Gene, or Tissue-Based Products. CAUTION: Not intended for direct administration into humans or animals.
Specifications
Cell TypeT cells
ClassificationAnimal Origin-free, Serum-free, cGMP
Culture TypeStatic Culture, Dynamic Bioreactors
FormLiquid
Serum LevelSerum-free
Shelf Life12 Months
SterilitySterile-filtered
Sterilization Method0.2 μm Sterile-filtered
With AdditivesNone
Without AdditivesNo Glutamine, No Phenol Red
FormatBPC
Product LineCTS™ OpTmizer™
Quantity5 L
Unit SizeEach
Contents & Storage
Store at 2-8°C and protect from light.