KingFisher™ Apex 96 Deep-well Head Magnetic Particle Processor for PrepSEQ Chemistry
KingFisher™ Apex 96 Deep-well Head Magnetic Particle Processor for PrepSEQ Chemistry
Thermo Scientific™
KingFisher™ Apex 96 Deep-well Head Magnetic Particle Processor for PrepSEQ Chemistry
The Thermo Scientific Pharma KingFisher Apex 96 Deep-well Head Magnetic Particle Processor is intended for pharmaceutical analytics applications and isRead more
Have Questions?
Catalog number A57715
Price (USD)
-
Request A Quote
The Thermo Scientific Pharma KingFisher Apex 96 Deep-well Head Magnetic Particle Processor is intended for pharmaceutical analytics applications and is an excellent option for facilities that wish to automate their PrepSEQ Residual DNA or PrepSEQ 1-2-3 nucleic acid extraction kit protocols. The KingFisher Apex platform is a premier automated platform designed to meet your high-throughput needs. It seamlessly incorporates the rapid, reliable, and cost-effective magnetic bead-based extraction of nucleic acids that you expect from PrepSEQ technology with a 96-well automation system. Using magnetic particle separation technology, this system provides excellent reproducibility and quality.
Features of the Pharma KingFisher Apex 96 Deep-well Head Magnetic Particle Processor include:
Reliable recovery—quantitative and consistent nucleic acid recovery and cross-contamination control
Efficient isolation—high-quality and high-throughput isolation of nucleic acids from Mycoplasma, Vesivirus, MMV, and residual DNA
Streamlined design—superior performance and cost effectiveness per reaction
Modular & versatile application—fully compatible with the PrepSEQ nucleic acid extraction kits
Quantitative & consistent recovery
Quantitative and consistent recovery of DNA from challenging matrices—including samples that have high protein concentration, low pH, or high salt—often presents a technical challenge. The PrepSEQ nucleic acid extraction kits are now adapted for higher-throughput workflows (up to 96 extractions, or 24 samples in triplicate) when used in combination with the KingFisher Flex 96 Deep-well Head Magnetic Particle Processor. The special formulation of magnetic particles provided in the PrepSEQ kits help ensure efficient and specific recovery of DNA from challenging samples.
High quality & throughput isolation
Most automated liquid handling systems move reagents into and out of a single well to perform the different steps of nucleic acid isolation. In contrast, the KingFisher Apex 96 Deep-well Head Magnetic Particle Processor utilizes permanent rods to collect the magnetic beads from solution and releases the beads into a well containing reagents for the next step of isolation. The effectiveness of bead collection and transfer leads to superior washing, elution efficiency, and processing. The KingFisher Apex 96 Deep-well Head Magnetic Particle Processor can process up to 24 samples in triplicate, compared to 3 samples in triplicate using the manual method.
Compatible with PrepSEQ sample preparation kits
The KingFisher Apex 96 Deep-well Head Magnetic Particle Processor has been optimized to work with both the PrepSEQ Residual DNA Sample Preparation Kit and the PrepSEQ 1-2-3 Nucleic Acid Extraction Kit assays. The instrument comes pre-installed with PrepSEQ optimized protocols and does not require any further optimization after installation and validation.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product TypeNucleic Acid Extraction and Cell Separation Instrument
Sample TypeTissue Homogenate
Suitable Downstream AnalysisPCR/qPCR, Sequencing, Mass Spec
Plates per Deck8
Format96-well Instrument
Unit SizeCase of 1
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link