Thermo Scientific™

Zeba™ Spin Desalting Columns and Plates, 40K MWCO, 75 μL–10 mL

Catalog number: A57759
Have Questions?
Thermo Scientific™

Zeba™ Spin Desalting Columns and Plates, 40K MWCO, 75 μL–10 mL

Catalog number: A57759
Have Questions?
The new and enhanced Thermo Scientific Zeba Spin and Micro Spin Desalting columns and plates are specifically designed for efficient desalting and buffer exchanging proteins with a molecular weight >40 kDa and removing small molecules less than 2 kDa. They contain a proprietary and enhanced size-exclusion chromatography resin with a more robust bead structure, making them ideal for handling centrifugal force pressure. The strengthened bead structure results in better flow characteristics and increased efficiency in removing low MW protein contaminants from samples.
 
Catalog Number
A57759
Unit Size
25 columns
Description
Spin Desalting Columns, 0.5 mL, 25 Columns
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionSpin Desalting Columns, 0.5 mL, 25 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)50 to 150 μL
Unit Size25 columns
Catalog Number
Unit Size
25 columns
Description
Micro Spin Desalting Columns, 25 Columns
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionMicro Spin Desalting Columns, 25 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)5 to 14 μL
Unit Size25 columns
Catalog Number
Unit Size
50 columns
Description
Micro Spin Desalting Columns, 50 Columns
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionMicro Spin Desalting Columns, 50 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)5 to 14 μL
Unit Size50 columns
Catalog Number
Unit Size
50 columns
Description
Spin Desalting Columns, 0.5 mL, 50 Columns
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionSpin Desalting Columns, 0.5 mL, 50 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)50 to 150 μL
Unit Size50 columns
Catalog Number
Unit Size
5 columns
Description
Spin Desalting Columns, 2 mL, 5 Columns
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionSpin Desalting Columns, 2 mL, 5 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)300 to 800 μL
Unit Size5 columns
Catalog Number
Unit Size
25 columns
Description
Spin Desalting Columns, 2 mL, 25 Columns
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionSpin Desalting Columns, 2 mL, 25 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)300 to 800 μL
Unit Size25 columns
Catalog Number
Unit Size
5 columns
Description
Spin Desalting Columns, 5 mL, 5 Columns
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionSpin Desalting Columns, 5 mL, 5 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)500–2000 μL
Unit Size5 columns
Catalog Number
Unit Size
25 columns
Description
Spin Desalting Columns, 5 mL, 25 Columns
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionSpin Desalting Columns, 5 mL, 25 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)500–2000 μL
Unit Size25 columns
Catalog Number
Unit Size
5 columns
Description
Spin Desalting Columns, 10 mL, 5 Columns
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionSpin Desalting Columns, 10 mL, 5 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)1000 to 4000 μL
Unit Size5 columns
Catalog Number
Unit Size
25 columns
Description
Spin Desalting Columns, 10 mL, 25 Columns
Price (USD)
Availability
-
Full specifications
Quantity
DescriptionSpin Desalting Columns, 10 mL, 25 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)1000 to 4000 μL
Unit Size25 columns
Showing 10 of 12Show more
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)AvailabilityQuantity
A57759Full specifications
25 columnsSpin Desalting Columns, 0.5 mL, 25 Columns
-
DescriptionSpin Desalting Columns, 0.5 mL, 25 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)50 to 150 μL
Unit Size25 columns
A57756Full specifications
25 columnsMicro Spin Desalting Columns, 25 Columns
-
DescriptionMicro Spin Desalting Columns, 25 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)5 to 14 μL
Unit Size25 columns
A57758Full specifications
50 columnsMicro Spin Desalting Columns, 50 Columns
-
DescriptionMicro Spin Desalting Columns, 50 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)5 to 14 μL
Unit Size50 columns
A57760Full specifications
50 columnsSpin Desalting Columns, 0.5 mL, 50 Columns
-
DescriptionSpin Desalting Columns, 0.5 mL, 50 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)50 to 150 μL
Unit Size50 columns
A57761Full specifications
5 columnsSpin Desalting Columns, 2 mL, 5 Columns
-
DescriptionSpin Desalting Columns, 2 mL, 5 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)300 to 800 μL
Unit Size5 columns
A57762Full specifications
25 columnsSpin Desalting Columns, 2 mL, 25 Columns
-
DescriptionSpin Desalting Columns, 2 mL, 25 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)300 to 800 μL
Unit Size25 columns
A57763Full specifications
5 columnsSpin Desalting Columns, 5 mL, 5 Columns
-
DescriptionSpin Desalting Columns, 5 mL, 5 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)500–2000 μL
Unit Size5 columns
A57764Full specifications
25 columnsSpin Desalting Columns, 5 mL, 25 Columns
-
DescriptionSpin Desalting Columns, 5 mL, 25 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)500–2000 μL
Unit Size25 columns
A57765Full specifications
5 columnsSpin Desalting Columns, 10 mL, 5 Columns
-
DescriptionSpin Desalting Columns, 10 mL, 5 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)1000 to 4000 μL
Unit Size5 columns
A57766Full specifications
25 columnsSpin Desalting Columns, 10 mL, 25 Columns
-
DescriptionSpin Desalting Columns, 10 mL, 25 Columns
Product TypeSpin Desalting Column
FormatSpin Column
Purification TargetBuffer Exchange, Protein
Column TypeSize-exclusion, Proprietary Resin
MWCO40 kDa
Product LineZeba™
DisposableSingle-use
Sample Volume (Metric)1000 to 4000 μL
Unit Size25 columns
Showing 10 of 12Show more

Separating macromolecules, such as proteins from small contaminants, is often done through routine methods including gel filtration chromatography and buffer exchange. These methods are commonly used during protein purification workflows to remove small contaminants like salts, excess labels, and cross-linking reagents from samples before analysis.

The new and enhanced Thermo Scientific Zeba Spin Desalting columns and plates are specifically designed for efficient desalting and buffer exchanging proteins with a molecular weight >40 kDa and removing small molecules less than 2 kDa. They contain a proprietary and enhanced size-exclusion chromatography resin with a more robust bead structure, making them ideal for handling centrifugal force pressure. The strengthened bead structure results in better flow characteristics and increased efficiency in removing low MW protein contaminants from samples. The Zeba resin performs exceptionally well in purifying protein samples, even when they are significantly diluted. It eliminates over 95% of contaminants without causing protein loss.

Zeba desalting columns are the ideal solution for quick and straightforward desalting and buffer exchanging of protein samples. They are offered in variousformats, such as pre-dispensed spin columns (75 μL, 0.5 mL, 2 mL, 5 mL, 10 mL), 96-well filter plates, and chromatography cartridges. These options make it simple to handle sample volumes between 2 μL and 4 mL.

Zeba Spin Desalting 96-well Plates provide trouble-free high throughput desalting and buffer exchange for sample volumes ranging from 20 to 100 μL. They require no resin dispensing or hydration, and provide the same high protein recovery as spin columns with at least 95% removal of salts and other small molecules. One plate can process 96 small sample volumes (20 to 100 μL) in five minutes.

Features and Benefits
High-performing resin—exceptional protein recovery at lower protein concentrations
Fast and Efficient—eliminate sample dilution, fraction collection, and the need for expensive chromatography system that take time to set up and can only process one sample at a time
Easy to use—no cumbersome column preparation or equilibration
Flexible formats—available in spin columns and 96-well spin plates for a range of sample volumes (2 μL to 4 mL)

For Research Use Only. Not for use in diagnostic procedures.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text