DNase/RNase-Free Deionized Water
DNase/RNase-Free Deionized Water
Thermo Scientific™
DNase/RNase-Free Deionized Water
DNase/RNase-Free Deionized Water for use in all molecular biology applications.
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2 L
Catalog number A57775
Price (USD)
70.25
Each
Add to cart
Quantity:
1 L
Price (USD)
70.25
Each
Add to cart
DNase/RNase-Free Deionized Water is designed for use in all molecular biology applications and provided in a convenient 1-liter and 2-liter bottles.
Confidence in your applications
The water is 0.22-micron-filtered to remove bacteria, most viruses, and other particulates, making it suitable for preparation of growth media. Water quality is monitored with extensive testing for the presence of DNA/RNA nucleases, protease activity, and human and bacterial DNA.
Applications
DNase/RNase-Free Deionized Water is suitable for various molecular biology applications, including PCR, qPCR, cDNA synthesis, cloning, next-generation sequencing, nucleic acid extraction or storage, and research applications involving low amounts of nucleic acids. Additionally, it can be used for applications that are sensitive to the presence of ions, like solution or buffer preparation, mass spectrometry, and spectrophotometry.
Manufacturing excellence
Manufactured by Thermo Fisher Scientific Baltics UAB in compliance with ISO 9001 and ISO 13485 certified quality management system.
For Research Use Only.
Specifications
SynonymNuclease-free deionized water
Treatment(s)Not DEPC-Treated
GradeMolecular Biology
Quantity1 L
Unit SizeEach
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link