1x1 image pixel for data collection
Pierce™ Crossflow Diafiltration Reservoir, 500 mL
Pierce™ Crossflow Diafiltration Reservoir, 500 mL
Thermo Scientific™
Pierce™ Crossflow Diafiltration Reservoir, 500 mL
Pierce Crossflow Diafiltration Reservoir, for continuous buffer replacement using Pierce Cross Flow Filtration Cassettes.
Have Questions?
Catalog NumberQuantity
A57935Each
Catalog number A57935
Price (USD)
240.00
Each
In stock
Add to cart
Quantity:
Each
Price (USD)
240.00
Each
Add to cart
The Pierce Crossflow Diafiltration Reservoir allows for continuous buffer replacement using Pierce Cross Flow Filtration Cassettes.
• For continuous buffer replacement using Pierce Cross Flow Filtration Cassettes.
Specifications
ColorClear
Includes3 ports
QuantityEach
AutoclavableNot Autoclavable
Bottom ShapeFlat
DisposableReusable
MaterialPlastic
SterilityNon-sterile
TypeSelf-standing Reagent Reservoir with Lid
Unit SizeEach
Frequently asked questions (FAQs)
Are Pierce Crossflow Filtration Cassettes GMP grade?
Pierce Crossflow Filtration Cassettes are not GMP grade.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Can I run Pierce Crossflow Filtration Cassettes without the pressure gauge?
A pressure gauge is provided with every Pierce Crossflow Filtration Cassette. Pierce Crossflow Filtration Cassettes can be run without a pressure gauge; however, it is important to limit the flow rate to 200-300 mL/min to prevent tearing and potential collapse of the tubing. The flow rate can be determined by measuring the volume flow over 1 min. Alternatively, a pump speed of 300-350 rpm or a pressure of 1-1.5 bar will maintain a flow rate of 200-300 mL/min.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What are the factors that can speed up or slow down the rate of concentration or diafiltration?
High protein concentration and/or presence of particulate in the sample can slow down the rate of concentration/diafiltration. Adsorption of the sample to the membrane and lower temperature (4 degrees C vs room temperature) can also slow down concentration/diafiltration. Filtering your sample prior to processing with a crossflow filtration cassette will help speed up concentration/diafiltration.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
What are the advantages of using a crossflow ultrafiltration device vs. a stirred cell device?
Crossflow ultrafiltration and stirred cell devices both effectively concentrate biomolecules with similar sample recoveries; however, a crossflow filtration device concentrates sample much faster than a stirred cell device.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Why should I use crossflow filtration instead of dialysis or chromatography methods?
Crossflow filtration is much faster for concentration and diafiltration than dialysis and chromatography methods.
Find additional tips, troubleshooting help, and resources within our Protein Purification and Isolation Support Center.
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link