Specifications
Bead TypeMagnetic
ColorBrown
FormatBottle
Isolation TechnologyBeads
Sample TypeSwabs in Transport Media, Saliva, Urine, Serum, Plasma
For Use With (Application)Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput CompatibilityHigh-throughput Compatible, Automated Protocols
Key FunctionsIsolate and purify viral, bacterial, parasitic, and fungal nucleic acid
No. of ReactionsUp to 2000 Reactions
Nucleic Acid TypeDNA/RNA
Product LineMagMAX™
Product TypeReagent
Sample Volume200–400 μL
Shipping ConditionRoom Temperature
Shelf Life2 Year
TargetViral, Bacterial, Parasitic, and Fungal Nucleic Acid
Unit Size20 mL