MagMAX™ Prime Viral/Pathogen Binding Solution
MagMAX™ Prime Viral/Pathogen Binding Solution
Applied Biosystems™

MagMAX™ Prime Viral/Pathogen Binding Solution

MagMAX Prime Viral/Pathogen Binding Solution is offered separately here for applications that require more Binding Solution than is provided inRead more
Have Questions?
Catalog number A58150
Price (USD)/ 530 mL
589.90
Special Offer
Online exclusive
Ends: 16-Dec-2024
694.00 
Save 104.10 (15%)
-
Add to cart
Price (USD)/ 530 mL
589.90
Special Offer
Online exclusive
Ends: 16-Dec-2024
694.00 
Save 104.10 (15%)
Add to cart
MagMAX™ Prime Viral/Pathogen Binding Solution
Catalog numberA58150
Price (USD)/ 530 mL
589.90
Special Offer
Online exclusive
Ends: 16-Dec-2024
694.00 
Save 104.10 (15%)
-
Add to cart

MagMAX Prime Viral/Pathogen Binding Solution is offered separately here for applications that require more Binding Solution than is provided in the MagMAX Prime Viral/Pathogen NA Isolation kits.

For Research Use Only.
Specifications
Bead TypeMagnetic
ColorClear
FormatBottle
Isolation TechnologyBeads
Sample TypeSwabs in Transport Media, Saliva, Urine, Serum, Plasma
For Use With (Application)Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput CompatibilityHigh-throughput Compatible, Automated Protocols
Key FunctionsIsolate and purify viral, bacterial, parasitic, and fungal nucleic acid
No. of ReactionsUp to 2000 Reactions
Nucleic Acid TypeDNA/RNA
Product LineMagMAX™
Product TypeReagent
Quantity530 mL
Sample Volume200–400 μL
TargetViral, Bacterial, Parasitic, and Fungal Nucleic Acid
Unit Size530 mL
Contents & Storage
Store at 15–25°C.