MagMAX™ Prime Viral/Pathogen Proteinase K
MagMAX™ Prime Viral/Pathogen Proteinase K
Applied Biosystems™

MagMAX™ Prime Viral/Pathogen Proteinase K

MagMAX Prime Viral/Pathogen Proteinase K is offered separately here for applications that require more Proteinase K than is provided inRead more
Have Questions?
Catalog number A58151
Price (USD)/ 10 mL
657.90
Special Offer
Online exclusive
Ends: 16-Dec-2024
774.00 
Save 116.10 (15%)
-
Add to cart
Price (USD)/ 10 mL
657.90
Special Offer
Online exclusive
Ends: 16-Dec-2024
774.00 
Save 116.10 (15%)
Add to cart
MagMAX™ Prime Viral/Pathogen Proteinase K
Catalog numberA58151
Price (USD)/ 10 mL
657.90
Special Offer
Online exclusive
Ends: 16-Dec-2024
774.00 
Save 116.10 (15%)
-
Add to cart

MagMAX Prime Viral/Pathogen Proteinase K is offered separately here for applications that require more Proteinase K than is provided in the MagMAX Prime Viral/Pathogen NA Isolation kits.

For Research Use Only.
Specifications
Bead TypeMagnetic
ColorClear
FormatBottle
Isolation TechnologyBeads
Sample TypeSwabs in Transport Media, Saliva, Urine, Serum, Plasma
For Use With (Application)Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput CompatibilityHigh-throughput Compatible, Automated Protocols
Key FunctionsIsolate and purify viral, bacterial, parasitic, and fungal nucleic acid
No. of ReactionsUp to 2000 Reactions
Nucleic Acid TypeDNA/RNA
Product LineMagMAX™
Product TypeReagent
Sample Volume200–400 μL
Shipping ConditionRoom Temperature
Shelf Life2 Years
TargetViral, Bacterial, Parasitic, and Fungal Nucleic Acid
Unit Size10 mL
Contents & Storage
Store at 15–25°C.