MagMAX™ Prime G+ Bacteria and Fungal Lysis Buffer
MagMAX™ Prime G+ Bacteria and Fungal Lysis Buffer
Applied Biosystems™

MagMAX™ Prime G+ Bacteria and Fungal Lysis Buffer

MagMAX Prime G+ Bacterial and Fungal Lysis Buffer is specifically designed to enable lysis and nucleic acid recovery from theRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
A58153Up to 1000 Reactions
A59053Up to 100 Reactions
2 Options
Catalog number A58153
Price (USD)/ 50 mL
2,388.50
Special Offer
Online exclusive
Ends: 16-Dec-2024
2810.00 
Save 421.50 (15%)
-
Add to cart
No. of Reactions:
Up to 1000 Reactions
Price (USD)/ 50 mL
2,388.50
Special Offer
Online exclusive
Ends: 16-Dec-2024
2810.00 
Save 421.50 (15%)
Add to cart
MagMAX™ Prime G+ Bacteria and Fungal Lysis Buffer
Catalog numberA58153
Price (USD)/ 50 mL
2,388.50
Special Offer
Online exclusive
Ends: 16-Dec-2024
2810.00 
Save 421.50 (15%)
-
Add to cart

MagMAX Prime G+ Bacterial and Fungal Lysis Buffer is specifically designed to enable lysis and nucleic acid recovery from the most challenging pathogens, such as gram-positive bacteria, yeast, and fungi. It is designed to be used in conjunction with MagMAX Prime Viral/Pathogen NA Isolation kits to maximize RNA and DNA yield.

For Research Use Only.
Specifications
Bead TypeMagnetic
ColorClear
FormatBottle
Isolation TechnologyBeads
Sample TypeSwabs in Transport Media, Saliva, Urine, Serum, Plasma
For Use With (Application)Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput CompatibilityHigh-throughput Compatible, Automated Protocols
Key FunctionsIsolate and purify viral, bacterial, parasitic, and fungal nucleic acid
No. of ReactionsUp to 1000 Reactions
Nucleic Acid TypeDNA/RNA
Product LineMagMAX™
Product TypeReagent
Sample Volume200–400 μL
Shipping ConditionDry Ice
TargetViral, Bacterial, Parasitic, and Fungal Nucleic Acid
Unit Size50 mL
Contents & Storage
Store at –15 to –25°C.