MagMAX™ Prime Bead Beating Tubes
MagMAX™ Prime Bead Beating Tubes
Applied Biosystems™

MagMAX™ Prime Bead Beating Tubes

MagMAX Prime Bead Beating Tubes are specifically designed to enable efficient, high-throughput nucleic acid extraction from stool. They are designedRead more
Have Questions?
Catalog NumberQuantity
A58155100 tubes
Catalog number A58155
Price (EUR)
192,00
Each
-
Add to cart
Quantity:
100 tubes
Recurring order eligible. Learn more »
Price (EUR)
192,00
Each
Add to cart
Ask our AI about this Product

MagMAX Prime Bead Beating Tubes are specifically designed to enable efficient, high-throughput nucleic acid extraction from stool. They are designed to be used with MagMAX Prime Stool Lysis Buffer and MagMAX Prime Viral/Pathogen NA Isolation kits to maximize RNA and DNA yield.

For Research Use Only.
Specifications
Bead TypeMagnetic
ColorClear
For Use With (Application)Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR
High-throughput CompatibilityHigh-throughput Compatible, Automated Protocols
Key FunctionsIsolate and purify viral, bacterial, parasitic, and fungal nucleic acid
No. of Reactions100 Reactions
Nucleic Acid TypeDNA/RNA
Product LineMagMAX™
Product TypeReagent
Quantity100 tubes
Sample TypeFecal Swab in Transport Media, Stool
Sample Volume400 μL
Shelf Life2 Years
Shipping ConditionRoom Temperature
TargetViral, Bacterial, Parasitic, and Fungal Nucleic Acid
FormatTube
Isolation TechnologyBeads
Unit SizeEach
Contents & Storage
Store at 15–25°C.
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
2405003Certificate of AnalysisJan 25, 2025A58155
1 result displayed, search above for a specific certificate

Safety Data Sheets

Other products to consider



Share catalog number, name or link

1x1 image pixel for data collection