Invitrogen™

ATTO-TAG™ CBQCA Derivatization Reagent (CBQCA; 3-(4-Carboxybenzoyl)quinoline-2-Carboxaldehyde)

Catalog number: A6222
Have Questions?
Invitrogen™

ATTO-TAG™ CBQCA Derivatization Reagent (CBQCA; 3-(4-Carboxybenzoyl)quinoline-2-Carboxaldehyde)

Catalog number: A6222
Have Questions?
Catalog Number
A6222
also known as A-6222
Unit Size
10 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A6222
also known as A-6222
10 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The ATTO-TAG™ CBQCA derivatization reagent (CBQCA; 3-(4carboxybenzoyl)quinoline- 2-carboxaldehyde) reacts specifically with primary amines to form conjugates that can be analyzed by electrophoretic or chromatographic methods. The resulting products of CBQCA are maximally excited at 450 nm or by the 442 nm spectral line of the He-Cd laser and have emission maxima at ∼550 nm. In capillary zone electrophoresis (CZE), the sensitivity of detection of ATTO-TAG™ CBQCA conjugates should be in the attomole range (10-18 moles). The high sensitivity, freedom from background and long-wavelength excitability make these potential reagents for researcher, diagnostic and forensic applications.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Chemical Reactivity
Amine
Reactive Moiety
Aldehyde
Shipping Condition
Room Temperature
Form
Solid
Product Line
ATTO-TAG™
Quantity
10 mg
For Use With (Application)
Cell Viability, Proliferation and Function; Protein Electrophoresis and Western Blotting
Target
Primary Amine
Type
Derivatization Kits and Reagents
Label or Dye
ATTO-TAG CBQCA

Contents & Storage

Store in freezer (-5 to -30°C) and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text