Invitrogen™

APTS (8-Aminopyrene-1,3,6-Trisulfonic Acid, Trisodium Salt)

Catalog number: A6257
Have Questions?
Invitrogen™

APTS (8-Aminopyrene-1,3,6-Trisulfonic Acid, Trisodium Salt)

Catalog number: A6257
Have Questions?
Catalog Number
A6257
also known as A-6257
Unit Size
10 mg
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
A6257
also known as A-6257
10 mg
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The aromatic amine of APTS (8-aminopyrene-1,3,6-trisulfonic acid, trisodium salt) can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3) to form new biotinylated probes.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Quantity
10 mg
Product Type
APTS
Chemical Reactivity
Ketone, Aldehyde
Shipping Condition
Room Temperature
Reactive Moiety
Amine, Primary Amine
Label or Dye
Pyrene
Label Type
Classic Dyes

Contents & Storage

Store at room temperature and protect from light.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text