Thermo Scientific™

CHAPSO (3-[(3-cholamidopropyl)dimethylammonio]-2-hydroxy-1-propanesulfonate)

Catalog number: A65501
Have Questions?
Thermo Scientific™

CHAPSO (3-[(3-cholamidopropyl)dimethylammonio]-2-hydroxy-1-propanesulfonate)

Catalog number: A65501
Have Questions?
Catalog Number
A65501
Unit Size
1 g
Price (USD)
Quantity
Catalog Number
Unit Size
5 g
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
A655011 g
A655025 g
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations

CHAPSO, or 3-[(3-cholamidopropyl)dimethylammonio]-2-hydroxy-1-propanesulfonate, is a zwitterionic detergent commonly used in biochemical and biophysical research. It is widely employed for membrane protein solubilization, stabilization, and purification. CHAPSO is known for its mildness and ability to preserve the structural integrity and functionality of membrane proteins during extraction. Its unique properties make it suitable for a variety of applications, including protein crystallization, spectroscopy, and structural biology studies. With its versatile and effective solubilization capabilities, CHAPSO is a valuable tool for researchers working with membrane proteins.

Features of CHAPSO include:
• Zwitterionic detergent
• Gentle—enabling preservation of native conformation and activity
• Effectively disrupts lipid-protein interactions
• Exhibits low critical micelle concentration (CMC)
• High-purity compound with low UV absorptivity

Properties
CMC: (H2O) ~8 mM (0.50%)
Aggregation number: (H2O) ~11
Purity: ≥99% (by HPLC analysis)
pH: 5–8 (1% solution in water)
Solubility: ≥0.5 M (in water at 20 ̊C)
Fluorescence: <10% (0.1% solution in water at 345 nm)
Absorbance of a 1% solution in water: 260 nm: <0.06, 280 nm: <0.04, 340 nm: <0.02

Specifications

Color
White
Product Type
Detergent
CAS
82473-24-3
Molecular Formula
C32H58N2O8S
Solubility
≥0.5 M (in water at 20°C)
Form
Lyophilized
Reagent Type
Detergent
Purification Target
Proteins
Quantity
1 g
Molecular Weight (g/mol)
630.9
Unit Size
1 g

Contents & Storage

Store below -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text