Thermo Scientific™

High-Purity Maltoside Detergents

Catalog number: A65510
Have Questions?
Thermo Scientific™

High-Purity Maltoside Detergents

Catalog number: A65510
Have Questions?
Maltoside detergents are a class of nonionic surfactants that are commonly utilized in the field of membrane protein solubilization. Several studies have shown that these surfactants are highly effective in extracting and stabilizing membrane proteins. Maltosides also aid in the purification and characterization of membrane proteins, facilitating downstream applications such as structural determination and functional analysis, making them an essential tool for researchers and scientists working in this area.
 
Catalog Number
A65510
Unit Size
Each
Description
n-Undecyl-β-D-maltopyranoside (UDM)
Quantity
5 g
Price (USD)
Availability
-
Full specifications
Quantity
ColorWhite
Descriptionn-Undecyl-β-D-maltopyranoside (UDM)
Product TypeDetergent
CAS253678-67-0
Molecular FormulaC23H44O11
Solubility≥20% (in water at 0°C to 5°C)
FormLyophilized
Quantity5 g
Molecular Weight (g/mol)496.6
pH5 to 8 (1% solution in water)
Unit SizeEach
Catalog Number
Unit Size
Each
Description
n-Dodecyl-β-D-Maltoside
Quantity
1 g
Price (USD)
Availability
-
Full specifications
Quantity
Descriptionn-Dodecyl-β-D-Maltoside
Product TypeDetergent
FormPowder
Quantity1 g
Molecular Weight (g/mol)510.6 g/mol
Unit SizeEach
Catalog Number
Unit Size
Each
Description
n-Dodecyl-β-D-Maltoside
Quantity
5 g
Price (USD)
Availability
-
Full specifications
Quantity
Descriptionn-Dodecyl-β-D-Maltoside
Product TypeDetergent
FormPowder
Quantity5 g
Molecular Weight (g/mol)510.6 g/mol
Unit SizeEach
Catalog Number
Unit Size
Each
Description
n-Undecyl-β-D-maltopyranoside (UDM)
Quantity
1 g
Price (USD)
Availability
-
Full specifications
Quantity
ColorWhite
Descriptionn-Undecyl-β-D-maltopyranoside (UDM)
Product TypeDetergent
CAS253678-67-0
Molecular FormulaC23H44O11
Solubility≥20% (in water at 0°C to 5°C)
FormLyophilized
Quantity1 g
Molecular Weight (g/mol)496.6
pH5 to 8 (1% solution in water)
Unit SizeEach
Catalog Number
Unit Size
Each
Description
Lauryl Maltose Neopentyl Glycol (LMNG)
Quantity
1 g
Price (USD)
Availability
-
Full specifications
Quantity
Chemical Name or Material2,2-didecylpropane-1,3-bis-β-D-maltopyranoside
ColorWhite
DescriptionLauryl Maltose Neopentyl Glycol (LMNG)
Molecular FormulaC47H88O22, C47H88O22
FormSolid
Molecular Weight (g/mol)1005.19
Quantity1 g
Percent Purity>98%
pH5–8 (1% solution in water)
Unit SizeEach
Catalog Number
Unit Size
Each
Description
LMNG/CHS Solution (10:1)
Quantity
10 x 1 mL
Price (USD)
Availability
-
Full specifications
Quantity
ColorColorless
DescriptionLMNG/CHS Solution (10:1)
FormLiquid
Quantity10 x 1 mL
Unit SizeEach
Catalog Number
Unit Size
Each
Description
n-Octyl-β-D-Maltopyranoside (OM)
Quantity
1 g
Price (USD)
Availability
-
Full specifications
Quantity
ColorWhite
Descriptionn-Octyl-β-D-Maltopyranoside (OM)
Product TypeDetergent
CAS82494-08-4
Molecular FormulaC20H38O11
Solubility≥20% (in water at 20°C)
FormLyophilized
Quantity1 g
Molecular Weight (g/mol)454.4
pH5 to 8 (1% solution in water)
Unit SizeEach
Catalog Number
Unit Size
Each
Description
n-Octyl-β-D-Maltopyranoside (OM)
Quantity
1 g
Price (USD)
Availability
-
Full specifications
Quantity
ColorWhite
Descriptionn-Octyl-β-D-Maltopyranoside (OM)
Product TypeDetergent
CAS82494-08-4
Molecular FormulaC20H38O11
Solubility≥20% (in water at 20°C)
FormLyophilized
Quantity1 g
Molecular Weight (g/mol)454.4
pH5 to 8 (1% solution in water)
Unit SizeEach
Catalog Number
Unit Size
Each
Description
n-Nonyl-Beta-Maltoside (NM)
Quantity
1 g
Price (USD)
Availability
-
Full specifications
Quantity
ColorWhite
Descriptionn-Nonyl-Beta-Maltoside (NM)
Product TypeDetergent
CAS106402-05-5
Molecular FormulaC21H40O11
Solubility≥20% (in water at 0°C to 5°C)
FormLyophilized
Quantity1 g
Molecular Weight (g/mol)468.5
pH5 to 8 (1% solution in water)
Unit SizeEach
Catalog Number
Unit Size
Each
Description
n-Nonyl-Beta-Maltoside (NM)
Quantity
5 g
Price (USD)
Availability
-
Full specifications
Quantity
ColorWhite
Descriptionn-Nonyl-Beta-Maltoside (NM)
Product TypeDetergent
CAS106402-05-5
Molecular FormulaC21H40O11
Solubility≥20% (in water at 0°C to 5°C)
FormLyophilized
Quantity5 g
Molecular Weight (g/mol)468.5
pH5 to 8 (1% solution in water)
Unit SizeEach
Showing 10 of 10
Catalog NumberSpecificationsUnit SizeDescriptionQuantityPrice (USD)AvailabilityQuantity
A65510Full specifications
Eachn-Undecyl-β-D-maltopyranoside (UDM)5 g
-
ColorWhite
Descriptionn-Undecyl-β-D-maltopyranoside (UDM)
Product TypeDetergent
CAS253678-67-0
Molecular FormulaC23H44O11
Solubility≥20% (in water at 0°C to 5°C)
FormLyophilized
Quantity5 g
Molecular Weight (g/mol)496.6
pH5 to 8 (1% solution in water)
Unit SizeEach
89902Full specifications
Each n-Dodecyl-β-D-Maltoside1 g
-
Descriptionn-Dodecyl-β-D-Maltoside
Product TypeDetergent
FormPowder
Quantity1 g
Molecular Weight (g/mol)510.6 g/mol
Unit SizeEach
89903Full specifications
Each n-Dodecyl-β-D-Maltoside5 g
-
Descriptionn-Dodecyl-β-D-Maltoside
Product TypeDetergent
FormPowder
Quantity5 g
Molecular Weight (g/mol)510.6 g/mol
Unit SizeEach
A65509Full specifications
Eachn-Undecyl-β-D-maltopyranoside (UDM)1 g
-
ColorWhite
Descriptionn-Undecyl-β-D-maltopyranoside (UDM)
Product TypeDetergent
CAS253678-67-0
Molecular FormulaC23H44O11
Solubility≥20% (in water at 0°C to 5°C)
FormLyophilized
Quantity1 g
Molecular Weight (g/mol)496.6
pH5 to 8 (1% solution in water)
Unit SizeEach
A50940Full specifications
Each Lauryl Maltose Neopentyl Glycol (LMNG)1 g
-
Chemical Name or Material2,2-didecylpropane-1,3-bis-β-D-maltopyranoside
ColorWhite
DescriptionLauryl Maltose Neopentyl Glycol (LMNG)
Molecular FormulaC47H88O22, C47H88O22
FormSolid
Molecular Weight (g/mol)1005.19
Quantity1 g
Percent Purity>98%
pH5–8 (1% solution in water)
Unit SizeEach
A50941Full specifications
Each LMNG/CHS Solution (10:1)10 x 1 mL
-
ColorColorless
DescriptionLMNG/CHS Solution (10:1)
FormLiquid
Quantity10 x 1 mL
Unit SizeEach
A65511Full specifications
Eachn-Octyl-β-D-Maltopyranoside (OM)1 g
-
ColorWhite
Descriptionn-Octyl-β-D-Maltopyranoside (OM)
Product TypeDetergent
CAS82494-08-4
Molecular FormulaC20H38O11
Solubility≥20% (in water at 20°C)
FormLyophilized
Quantity1 g
Molecular Weight (g/mol)454.4
pH5 to 8 (1% solution in water)
Unit SizeEach
A65512Full specifications
Eachn-Octyl-β-D-Maltopyranoside (OM)1 g
-
ColorWhite
Descriptionn-Octyl-β-D-Maltopyranoside (OM)
Product TypeDetergent
CAS82494-08-4
Molecular FormulaC20H38O11
Solubility≥20% (in water at 20°C)
FormLyophilized
Quantity1 g
Molecular Weight (g/mol)454.4
pH5 to 8 (1% solution in water)
Unit SizeEach
A65513Full specifications
Eachn-Nonyl-Beta-Maltoside (NM)1 g
-
ColorWhite
Descriptionn-Nonyl-Beta-Maltoside (NM)
Product TypeDetergent
CAS106402-05-5
Molecular FormulaC21H40O11
Solubility≥20% (in water at 0°C to 5°C)
FormLyophilized
Quantity1 g
Molecular Weight (g/mol)468.5
pH5 to 8 (1% solution in water)
Unit SizeEach
A65514Full specifications
Eachn-Nonyl-Beta-Maltoside (NM)5 g
-
ColorWhite
Descriptionn-Nonyl-Beta-Maltoside (NM)
Product TypeDetergent
CAS106402-05-5
Molecular FormulaC21H40O11
Solubility≥20% (in water at 0°C to 5°C)
FormLyophilized
Quantity5 g
Molecular Weight (g/mol)468.5
pH5 to 8 (1% solution in water)
Unit SizeEach
Showing 10 of 10

Features of maltoside detergents
• Lipid-like nonionic detergent
• Especially useful for isolating and stabilizing hydrophobic membrane proteins
• Preserves activity of membrane protein better than most of the detergents
• High-purity compounds with low UV absorptivity

n-Dodecyl-β-D-Maltoside
n-Dodecyl-β-D-Maltoside (DDM) is especially useful for solubilizing membrane proteins to preserve their activity. This water-soluble nonionic detergent is most often used for the isolation of hydrophobic membrane proteins. Multiple studies have shown that n-dodecyl-β-D-maltoside is a gentle detergent that is often able to preserve protein activity better than many commonly used detergents, including NP-40, CHAPS, and octyl-β-glucoside.

Properties of DDM
Aggregation number: 98 (average), 70 to 140 range
Micelle molecular weight: 50,000 g
CMC: 0.17 mM (0.009%, w/v) in water ; 0.12 mM (0.006%, w/v) in 0.2 M NaCl

DDM/CHS Solution
DDM/CHS Solution (10:1) is a ready-to-use solution for solubilization of membrane proteins while maintaining structural integrity and activity. The stable pre-formulated solution contains a mixture of dodecyl-β-D-maltoside (DDM) (100 mg/mL) and cholesteryl hemisuccinate (CHS) (10 mg/mL) in deionized water with final concentrations of 10% and 1%, respectively. Typical working concentrations are 1% DDM/0.1% CHS for protein extraction and 0.1% DDM/0.01% CHS for protein purification. The mixture of nonionic detergent, DDM, with dual hydrophobic/hydrophilic properties and anionic detergent, CHS, aid in the stabilization of large unilamellar vesicles and is suitable for protein solubilization and stabilization for single particle cryo-EM analysis.

Additional properties of CHS
• Molecular formula: C31H50O4 · C4H11NO3
• Molecular weight: 607.9 g/mol
• Detergent: ionizable anionic
• Function:  stabilizes large unilamellar vesicles

Lauryl Maltose Neopentyl Glycol
Lauryl Maltose Neopentyl Glycol (LMNG) is an amphiphilic detergent ideally suited to improve solubilization of integral membrane proteins while maintaining structural integrity and activity.
LMNG has two linked hydrophobic chains of equal length and two hydrophilic maltoside groups to better mimic the integral membrane protein environment. Micelles are known to aid in solubilization of membrane proteins while preserving structure and function, including G-protein coupled receptors (GPCRs). LMNG is suitable for protein solubilization and stabilization for single particle cryo-EM analysis.

Properties of LMNG
• Chemical name: 2,2-didecylpropane-1,3-bis-β-D-maltopyranoside
• Molecular weight: 1005.19 g/mol
• CMC: (H2O) ∼ 0.01 mM, (0.001%)
• Purity: ≥98% all anomers (by HPLC analysis)

LMNG/CHS Solution
LMNG/CHS Solution (10:1) is a ready-to-use formulation for the solubilization of integral membrane proteins while maintaining structural integrity and activity. The stable pre-formulated solution contains a mixture of lauryl maltose neopentyl glycol (LMNG) (5%) and cholesteryl hemisuccinate (CHS) (0.5%) in deionized water. Typical working concentrations for protein extraction are 1% LMNG/0.1% CHS and 0.01% LMNG/0.001% CHS for protein purification and structure determination. LMNG/CHS micelles aid in solubilization of membrane proteins while preserving structure and function, including G-protein coupled receptors (GPCRs), and is suitable for protein solubilization and stabilization for single particle cryo-EM analysis.

n-Undecyl-β-D-Maltopyranoside
n-Undecyl-β-D-Maltopyranoside (UDM) has an undecyl (C11) alkyl chain attached to the maltose headgroup. The longer alkyl chain length of UDM contributes to its enhanced solubilization efficiency for certain membrane proteins. The increased hydrophobicity of the undecyl chain may facilitate stronger interactions with hydrophobic regions of membrane proteins, leading to improved solubilization and stabilization. The longer undecyl chain length in UDM may also result in a higher CMC compared to maltosides with shorter alkyl chains. This can affect the formation and stability of micelles in solution.

Properties of UDM
CMC: (H2O) ~0.59 mM(1) (0.029%)
Aggregation number: ~71 (100 mM NaCl, 20 mM HEPES pH 7.5)
Purity: ≥99% β+α (by HPLC analysis)

n-Nonyl-Beta-Maltoside
n-Nonyl-Beta-Maltoside (NM) has a nonyl (C9) alkyl chain attached to the maltose headgroup. This specific chain length provides NM with distinct solubilization and stabilization properties compared to other maltosides with different alkyl chain lengths, such as octyl or decyl maltoside. The specific alkyl chain length of NM influences its micelle properties and contributes to a relatively low CMC. This property allows for the formation of stable micelles at lower concentrations, important for maintaining the stability and integrity of membrane proteins during solubilization and subsequent experiments.

Properties of NM
CMC: (H2O) ~ 6 mM (0.28%)
Aggregation number: ~55 (100 mM NaCl, 20 mM HEPES pH 7.5)
Purity: ≥99% β+α (by HPLC analysis)

n-Octyl-β-D-Maltopyranoside
n-Octyl-β-D-Maltopyranoside (OM) has an octyl (C8) alkyl chain attached to the maltose headgroup. The specific alkyl chain length of OM sets it apart from other maltosides with different alkyl chain lengths, such as nonyl or decyl maltoside. The octyl chain length in OM contributes to a relatively low CMC, allowing for the formation of stable micelles at lower concentrations. This property is important for maintaining the stability and integrity of membrane proteins during solubilization and subsequent experiments. OM is versatile and finds applications in various areas of membrane protein research, protein crystallization, and biochemical studies. It can be used in a wide range of experimental conditions, buffer systems, and pH ranges. OM offers flexibility and compatibility with different experimental setups.

Properties of OM
CMC: (100 mM NaCl, 20 mM HEPES pH 7.5) ~ 19.5 mM(1) (0.89%)
Aggregation number: ~47 (100 mM NaCl, 20 mM HEPES pH 7.5)
Purity: ≥99% β+α (by HPLC analysis)

For Research Use Only.

Contents & Storage

Store below -20°C.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text