1x1 image pixel for data collection
Easy Pierce Heat Sealing Foil
Easy Pierce Heat Sealing Foil
Actual product may vary
Thermo Scientific™

Easy Pierce Heat Sealing Foil

Use this heat sealing foil to ready your microplate for compound management and high-throughput screening applications, such as PCR.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberMaterialProduct TypeDimensions (L x W)
AB-0559AluminumThermo-Seal85 x 135 mm
AB-1720AluminumEasyPierce 20μm Thin Foil85 x 135 mm
AB-0757AluminumEasy Pierce Foil85 x 135 mm
AB-0812PolyesterDiamond Clear Seal85 x 135 mm
AB-1797PolyesterClear Seal Thin80 x 125 mm
AB-0745AluminumEasy Peel Film Laminate85 x 135 mm
Catalog number AB-0559
Price (USD)
126.54
Online Exclusive
153.00
Save 26.46 (17%)
Case of 100
Add to cart
Material:
Aluminum
Product Type:
Thermo-Seal
Dimensions (L x W):
85 x 135 mm
Price (USD)
126.54
Online Exclusive
153.00
Save 26.46 (17%)
Case of 100
Add to cart

Seal your microplate for compound management and high-throughput screening, including PCR, with Thermo Scientific™ Easy Pierce Heat Sealing Foil. Suitable for use with Dimethyl sulfoxide (DMSO) reagents, this sealing foil creates a peelable bond that can be easily pierced and removed by hand.

  • Suitable for PCR (requires the pressure from a screw-down heated lid)
  • Easily pierced
  • Peelable bond can be removed by hand
  • 20μm foil is even more easily pierced and is suitable for multiple layering
  • Recommended for use with DMSO
  • Seal Integrity Range: -80°C to +80°C (+120°C with adequate pressure from cycler lid)

Specifications
Dimensions (L x W)85 x 135 mm
For Use With (Application)Transportation and high temperature applications such as PCR
For Use With (Equipment)ALPS 25 and ALPS 50V; AB-1443A
Incubation ConditionsGeneral
No. per Case100
No. per Pack100
PeelableVery Strong Seal that requires stripper tool for peeling
Product TypeThermo-Seal
AdhesivePolypropylene
AutoclavableNot Autoclavable
Certified RNase/DNase-freeNo
ColorFoil
MaterialAluminum
No. of Wells96
PierceableYes
Plate CompatibilityPolypropylene
Product LineAbgene
Solvent Or Alcohol ResistantLife Science Reagents Resistant, DMSO Resistant
SterilityNon-sterile
Temperature Range (Metric)-80°C to +80°C, -80°C to +120°C (with Adequate Pressure)
Well ShapeAll well shapes
Unit SizeCase of 100
Contents & Storage
General; Long term storage including storage at low temperatures

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link