Thermo Scientific™
Abgene™ Multisip™ Plugs
Catalog number: AB-0972
Have Questions?
Thermo Scientific™
Abgene™ Multisip™ Plugs
Catalog number: AB-0972
Have Questions?
Thermo Scientific™ Abgene Multisip Plugs are made of an advanced thermoplastic elastomer that offers excellent chemical resistance and low vapor transmission rates, minimizing evaporationand water uptake in DMSO-based samples.The plug’s V-shaped design reliably guides liquid handling needles to the same location repeatedly, while this elastomer ensures sealing performance will not degrade, even after hundreds of cap piercings.
Catalog Number
AB-0972
Unit Size
Each
Description
Multisip Split Septum Plugs, 7 mm, Bulk
Price (USD)
Full specifications
Diameter (Metric)7 mm
DescriptionMultisip Split Septum Plugs, 7 mm, Bulk
AutoclavableAutoclavable
MaterialThermoplastic Elastomer
SterilityNon-sterile
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
AB-0972Full specifications
EachMultisip Split Septum Plugs, 7 mm, Bulk
Diameter (Metric)7 mm
DescriptionMultisip Split Septum Plugs, 7 mm, Bulk
AutoclavableAutoclavable
MaterialThermoplastic Elastomer
SterilityNon-sterile
Unit SizeEach
Showing 1 of 1
Features:
  • Enhanced Materials
  • Application Flexibility