Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Save time and costs while improving sealing integrity with Thermo Scientific™ Heat Seals for ALPS™ Microplate Heat Sealing Instruments. Ideal for storage applications, heat sealing offers the capability of retrieving samples and then re-sealing the plate. When heat sealing, the sealing material is first placed in position over the plate, which must have a raised rim around each well. Heat is applied evenly for several seconds, resulting in binding of the sealing material to the raised rim to achieve a complete seal.
Color | Foil |
Solvent Or Alcohol Resistant | Life Science Reagents Resistant, DMSO Resistant |
Description | Easy Pierce Foil, Roll, 78 mm x 610 m |
Material | Aluminum |
Sterility | Non-sterile |
Autoclavable | Not Autoclavable |
No. of Wells | 96 |
No. per Case | 1 |
No. per Pack | 1 |
Dimensions (L x W) | 610 m x 78 mm |
For Use With (Application) | ideal for compound management and high throughput screening applications, suitable for PCR |
For Use With (Equipment) | ALPS 5000 & ALPS 3000 |
Product Type | Automation Heat Seal |
Product Line | Abgene |
Adhesive | Polypropylene |
Certified RNase/DNase-free | No |
Pierceable | Yes |
Temperature Range (Metric) | -80°C to +120°C |
Well Shape | All well shapes |
Plate Compatibility | Polypropylene, Polyethylene |
Incubation Conditions | General |
Peelable | Yes |
Unit Size | Each |
Color | Clear |
Description | Clear Seal Thin, Polymer, Roll 78 mm x 610 m |
Material | Polyester |
No. of Wells | 96 |
No. per Case | 1 |
No. per Pack | 1 |
Dimensions (L x W) | 610 m x 78 mm |
For Use With (Application) | Ideal for fluorescence, colorimetric, and optical applications; suitable for PCR/ qPCR; ELISA |
For Use With (Equipment) | ALPS 5000 & ALPS 3000 |
Product Type | Automation Heat Seal |
Product Line | Abgene |
Adhesive | Polypropylene |
Pierceable | Yes |
Temperature Range (Metric) | -80°C to +120°C |
Plate Compatibility | Polypropylene, Polyethylene, Polystyrene |
Incubation Conditions | General |
Peelable | Yes |
Fluorescence | Low Autofluorescence |
Unit Size | Each |
Color | Foil |
Solvent Or Alcohol Resistant | Life Science Reagents Resistant, DMSO Resistant |
Description | ThermoSeal Foil Laminate, Roll, 78 mm x 370 m |
Material | Aluminum |
Sterility | Non-sterile |
Autoclavable | Not Autoclavable |
No. of Wells | 96 |
No. per Case | 1 |
No. per Pack | 1 |
Dimensions (L x W) | 370 m x 78 mm |
For Use With (Application) | Transportation and high temperature applications such as PCR |
For Use With (Equipment) | ALPS 500 & ALPS 3000 |
Product Type | Automation Heat Seal |
Product Line | Abgene |
Adhesive | Polypropylene |
Certified RNase/DNase-free | No |
Pierceable | Yes |
Temperature Range (Metric) | -80°C to +120°C |
Well Shape | All well shapes |
Plate Compatibility | Polypropylene |
Incubation Conditions | General |
Peelable | Very Strong Seal that requires stripper tool for peeling |
Unit Size | Each |
Color | Clear |
Solvent Or Alcohol Resistant | Some Solvent Resistance |
Description | Diamond ClearSeal Polymer Roll, 78 mm x 370 m |
Material | Polyester |
No. per Case | 1 |
No. per Pack | 1 |
Dimensions (L x W) | 370 m x 78 mm |
For Use With (Application) | Ideal for qPCR and other fluorescence, colorimetric, and optical applications; suitable for PCR; ELISA |
For Use With (Equipment) | ALPS 5000 & ALPS 3000; Recommended for sealing Fast Diamond 384 PCR plates |
Product Type | Automation Heat Seal |
Product Line | Abgene |
Adhesive | Polypropylene |
Pierceable | No |
Temperature Range (Metric) | -80°C to +120°C |
Plate Compatibility | Polypropylene, Polyethylene, Polystyrene |
Incubation Conditions | General |
Peelable | Yes |
Fluorescence | Low Autofluorescence |
Unit Size | Each |
Color | Foil |
Solvent Or Alcohol Resistant | Life Science Reagents Resistant, DMSO Resistant |
Description | EasyPierce 20 μm Thin Foil, Roll, 78 mm x 610 m |
Material | Aluminum |
Sterility | Non-sterile |
Autoclavable | Not Autoclavable |
No. of Wells | 96 |
No. per Case | 1 |
No. per Pack | 1 |
Dimensions (L x W) | 610 m x 78 mm |
For Use With (Application) | Ideal for compound management and high throughput screening applications, suitable for PCR |
For Use With (Equipment) | ALPS5000 & ALPS 3000 |
Product Type | Automation Heat Seal |
Product Line | Abgene |
Adhesive | Polypropylene |
Certified RNase/DNase-free | No |
Pierceable | Yes |
Temperature Range (Metric) | -80°C to +120°C |
Well Shape | All well shapes |
Plate Compatibility | Polypropylene, Polyethylene |
Incubation Conditions | General |
Peelable | Yes |
Unit Size | Each |
Color | Foil |
Solvent Or Alcohol Resistant | Life Science Reagents Resistant, DMSO Resistant |
Description | Easy-Peel Foil Laminate, Roll, 78 mm x 610 m |
Material | Aluminum |
Sterility | Non-sterile |
Autoclavable | Not Autoclavable |
No. of Wells | 96 |
No. per Case | 1 |
No. per Pack | 1 |
Dimensions (L x W) | 610 m x 78 mm |
For Use With (Application) | Ideal for long term storage at low temperatures; PCR |
For Use With (Equipment) | ALPS 5000 & ALPS 3000 |
Product Type | Automation Heat Seal |
Product Line | Abgene |
Adhesive | Polypropylene, Forms weld seal with polyethylene plates, seal can be applied, removed and a new seal applied several times |
Certified RNase/DNase-free | No |
Pierceable | No |
Temperature Range (Metric) | -200°C to +90°C |
Well Shape | All well shapes |
Plate Compatibility | Polypropylene, Polyethylene, Cyclic Olefin Copolymer |
Incubation Conditions | General |
Peelable | Yes |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Dimensions (L x W) | Price (USD) | Availability | Quantity | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
AB-3738 | Each | Easy Pierce Foil, Roll, 78 mm x 610 m | 610 m x 78 mm | 2,470.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AB-3797 | Each | Clear Seal Thin, Polymer, Roll 78 mm x 610 m | 610 m x 78 mm | 1,235.00 Online price 2,470.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AB-3559 | Each | ThermoSeal Foil Laminate, Roll, 78 mm x 370 m | 370 m x 78 mm | 2,088.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AB-3799 | Each | Diamond ClearSeal Polymer Roll, 78 mm x 370 m | 370 m x 78 mm | 1,920.96 Online price 2,088.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AB-3720 | Each | EasyPierce 20 μm Thin Foil, Roll, 78 mm x 610 m | 610 m x 78 mm | 2,470.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AB-3739 | Each | Easy-Peel Foil Laminate, Roll, 78 mm x 610 m | 610 m x 78 mm | 2,285.65 Online price 2,470.00 | - | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Features: