1x1 image pixel for data collection
Tubes, strips of 8
Try our New Thermo Scientific PCR Plastics, Sustain Series - identical performance and quality with a lower carbon footprint
Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Tubes, strips of 8
Thermo Scientific™

Tubes, strips of 8

The Thermo Scientific 0.2 mL Strip Tubes are designed for PCR and qPCR applications, and are compatible with 0.2 mLRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorClosure TypeProduct TypeQuantity
AB0264NaturalNot Included8-Strip Tube250 Strips
AB0452NaturalNot Included8-Strip Tube10 Packs of 12 Strips
Catalog number AB0264
Price (USD)
256.65
Online Exclusive
317.00
Save 60.35 (19%)
Each
Add to cart
Color:
Natural
Closure Type:
Not Included
Product Type:
8-Strip Tube
Quantity:
250 Strips
Price (USD)
256.65
Online Exclusive
317.00
Save 60.35 (19%)
Each
Add to cart
The Thermo Scientific 0.2 mL Strip Tubes are designed for PCR and qPCR applications, and are compatible with 0.2 mL thermal cycler blocks.While these tubes are sold without caps, Thermo Scientific offers multiple cap options, including an ultra clear cap that's ideal for use in qPCR assays. The caps form a secure seal, yet are easy to apply and remove. Caps increase the overall tube capacity to 0.25 mL

Features of The Thermo Scientific 0.2 mL Strip Tubes without caps

• Compatible with 0.2 mL thermal cycler blocks
• 8 tube strips
• Sealable using flat and domed cap strips. Ultra-Clear cap options available for use in qPCR assays

Applications

• PCR
• qPCR/Real-Time PCR

Related Products
Tubes, strips of 8, white
Tubes, strips of 8, blue
Tubes, strips of 8, green
Tubes, strips of 8, purple
Tubes, strips of 8, red
Tubes, strips of 8, yellow
Tubes, strips of 8
Tubes, strips of 12
Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Equipment)Thermal Cyclers
Format250 Strip Tubes
Quantity250 Strips
Reaction SpeedStandard
Volume (Metric) Working0.2 mL
Closure TypeNot Included
ColorNatural
MaterialPolypropylene
Product Type8-Strip Tube
Volume (Metric)0.25 mL
Unit SizeEach

Frequently asked questions (FAQs)

Are Thermo Scientific 0.2 mL Strip Tubes autoclavable?

Yes, our Thermo Scientific PCR plastics are autoclavable. Our recommendation is to autoclave for 15 min at 121 degrees C.

Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Instruments Support Center.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link