Domed PCR Caps, strips of 8
Try our New Thermo Scientific PCR Plastics, Sustain Series - identical performance and quality with a lower carbon footprint
Domed PCR Caps, strips of 8
Thermo Scientific™
Domed PCR Caps, strips of 8
Thermo Scientific Domed PCR Cap Strips of 8, are compatible with 96-well PCR plates and standard and low profile stripRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityColor
AB0265250 stripsClear
AB038610 packs of 12Clear
Catalog number AB0265
Price (USD)
100.65
Online Exclusive
134.00
Save 33.35 (25%)
Each
Add to cart
Quantity:
250 strips
Color:
Clear
Price (USD)
100.65
Online Exclusive
134.00
Save 33.35 (25%)
Each
Add to cart
Thermo Scientific Domed PCR Cap Strips of 8, are compatible with 96-well PCR plates and standard and low profile strip tubes.
Features of Thermo Scientific Domed PCR Cap Strips
• Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
Related Products
Domed PCR Caps, strips of 8
Domed PCR Caps, strips of 8, blue
Domed PCR Caps, strips of 8, green
Domed PCR Caps, strips of 8, purple
Domed PCR Caps, strips of 8, red
Domed PCR Caps, strips of 8, yellow
Domed PCR Caps, strips of 8, natural
Domed PCR Caps, strips of 12
Specifications
FormatStrips of 8 Domed Caps
Product TypePCR Plate Cap Strip
Quantity250 strips
Certified RNase/DNase-freeYes
ColorClear
MaterialPolypropylene
No. of Wells8
Plate Compatibility96-well Plate
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Frequently asked questions (FAQs)
I would like to use the flat ultra-clear and domed cap strips for Thermo-Fast PCR Plate, 48-well, clear (Cat. No. AB0648). What do you recommend?
For the domed cap strips, you could choose the Domed PCR Caps, strips of 8 (Cat. No. AB0265). For the flat ultra clear cap strips you can choose for the flat ultra clear cap strips the (Cat. No. AB0866). Generally speaking, within the Thermo Scientific/ABgene legacy, all our cap strips are compatible with all PCR plates and tube strips. For more options, please look at the following link.
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.
Which caps are Thermo-Fast PCR Plate, 24-well (Cat. No. AB0624) compatible with?
This plate is compatible with all caps e.g. Flat PCR Caps, strips of 8 (Cat. No. AB0784) and Domed PCR Caps, strips of 8 (Cat. No. AB0265).
Find additional tips, troubleshooting help, and resources within our Real-Time PCR and Digital PCR Applications Support Center.
Can I order the Caps within the Low Profile Tubes (white) and Ultra Clear Caps, strips of 8 (Cat. No. AB1771) seperately?
Yes. The suitable stand alone products are either the Ultra Clear qPCR Caps, strips of 8 (Cat. No.AB0866) or the Domed PCR Caps, strips of 8 (Cat. No. AB0265).
Documents & Downloads
Certificates
Safety Data Sheets
Share catalog number, name or link