You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
Shop All Products
Microplates
Microplate Covers
Microplate Well Caps
Domed PCR Caps, strips of 8
Thermo Scientific™
Domed PCR Caps, strips of 8
Catalog number:
AB0386
Related applications:
PCR
Have Questions?
Contact Us
Thermo Scientific™
Domed PCR Caps, strips of 8
Catalog number:
AB0386
Related applications:
PCR
Have Questions?
Contact Us
Catalog Number
AB0386
Unit Size
120 strips
Price (USD)
44.65
Online price
60.25
Quantity
Save to list
Add to cart
Catalog Number
AB0265
Unit Size
250 strips
Price (USD)
97.65
Online price
124.00
Quantity
Save to list
Add to cart
Catalog Number
Unit Size
Price (USD)
Quantity
AB0386
120 strips
44.65
Online price
60.25
AB0265
250 strips
97.65
Online price
124.00
Save to list
Add to cart
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Domed PCR Cap Strips of 8, are compatible with 96-well PCR plates and standard and low profile strip tubes.
Features of Thermo Scientific Domed PCR Cap Strips
• Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
Related Products
Domed PCR Caps, strips of 8
Domed PCR Caps, strips of 8, blue
Domed PCR Caps, strips of 8, green
Domed PCR Caps, strips of 8, purple
Domed PCR Caps, strips of 8, red
Domed PCR Caps, strips of 8, yellow
Domed PCR Caps, strips of 8, natural
Domed PCR Caps, strips of 12
Specifications
Color
Clear
Material
Polypropylene
No. of Wells
8
Quantity
10 packs of 12
Format
Strips of 8 Domed Caps
Product Type
PCR Plate Cap Strip
Certified RNase/DNase-free
Yes
Plate Compatibility
96-well Plate
Figures
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Frequently asked questions (FAQs)
Citations & References
Search citations by name, author, journal title or abstract text
Search