Tubes and Domed Caps, strips of 8, assorted colors
Need help choosing the right PCR/qPCR plastics? Use our on-line selection tool »
Tubes and Domed Caps, strips of 8, assorted colors
Thermo Scientific™

Tubes and Domed Caps, strips of 8, assorted colors

Thermo Scientific Tubes and Domed Caps, Strips of 8 are designed for PCR applications, and are suitable for 0.2 mLRead more
Have Questions?
Catalog number AB0490
Price (USD)/ 250 strips
357.65
Online exclusive
432.00 
Save 74.35 (17%)
Add to cart
Price (USD)/ 250 strips
357.65
Online exclusive
432.00 
Save 74.35 (17%)
Add to cart
Tubes and Domed Caps, strips of 8, assorted colors
Catalog numberAB0490
Price (USD)/ 250 strips
357.65
Online exclusive
432.00 
Save 74.35 (17%)
-
Add to cart
Thermo Scientific Tubes and Domed Caps, Strips of 8 are designed for PCR applications, and are suitable for 0.2 mL thermal cycler blocks and 96-well V-bottom thermal cyclers. The 8-strip tubes can be divided into individual tubes.

Features of Thermo Scientific Tubes and Domed Caps strips, assorted:

• Suitable for 0.2 mL thermal cycler blocks
• 0.25 mL maximum tube capacity when capped
• 8 tube strips can be cut into individual tubes
• Ideal for use in 96-well V-bottom thermal cyclers
• Colored tubes and caps available for keeping track of samples

Applications

• PCR

Related Products

Tubes and Domed Caps, strips of 8
Tubes and Domed Caps, strips of 8, blue
Tubes and Domed Caps, strips of 8, green
Tubes and Domed Caps, strips of 8, purple
Tubes and Domed Caps, strips of 8, red
Tubes and Domed Caps, strips of 8, yellow
Specifications
Certifications/ComplianceCertified DNA/RNase Free
ColorAssorted
Volume (Metric) Working0.2 mL
Volume (Metric)0.25 mL
Quantity250 Strips
For Use With (Equipment)Thermal Cyclers
MaterialPolypropylene
Reaction SpeedStandard
Closure TypeDomed Caps
No. of Wells96
Format250 Strip Tubes and Caps
Product Type8-Strip Tube
Unit Size250 strips