Thermo Scientific™

Workstation/Stacking System for Tubes/Plates

Catalog number: AB0538
Have Questions?
Thermo Scientific™

Workstation/Stacking System for Tubes/Plates

Catalog number: AB0538
Have Questions?
Catalog Number
AB0538
Unit Size
10 units
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
AB053810 units
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Work Station Stacking System is a convenient platform for tube handling and reagent dispensing

Features of the The Thermo Scientific Work Station Stacking System

• 96 x 0.2ml V-bottom tubes or
• 96 x 0.3ml U-bottom Omnistrips or
• Thermo-Tube Plates or
• Thermo-Fast 96 Non-Skirted Plate or
• Thermo-Fast 96 Semi-Skirted Plate or
• 48 x 0.5ml Thermo-Tubes
• Multiple workstations can be securely stacked
• Contains 10 units
• Can be stored at -190°C

Specifications

For Use With (Equipment)
Real-Time PCR Systems or Thermal Cyclers

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text