Thermo Scientific™

Gene Frame, 65 μL (1.5 x 1.6 cm)

Catalog number: AB0577
Have Questions?
Thermo Scientific™

Gene Frame, 65 μL (1.5 x 1.6 cm)

Catalog number: AB0577
Have Questions?
Catalog Number
AB0577
Unit Size
100 frames
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
AB0577100 frames
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Gene Frames have adhesives on either side of the frame which are of differing strengths; the side which adheres to the glass has an easy release adhesive whereas the reverse, which sticks to the coverslip, forms a stronger bond. This means that when the coverslip is removed, due to the disparity in the strengths of the different adhesives, the frame is removed simultaneously with no sticky residue being left behind on the slide.

Features of Thermo Scientific Gene Frames:

• Gas tight sealing prevents reagent loss due to evaporation
• Can withstand temperatures of 97°C
• Designed for use with standard microscope slides
• Frame and coverslip can be removed simultaneously leaving no residue
• Can be autoclaved or sterilized by UV irradiation
• All Gene Frame packs are supplied with 100 coverslips
• Rigid coverslips for ease of handling and uniform hybridizations
• Thickness of Gene Frames: 0.25 mm
• The 65 μL size have a 1.5 x 1.6 cm chamber

Applications

• In-situ PCR

Related Products
Gene Frame, 25 μL (1.0 x 1.0 cm)
Gene Frame, 125 μL (1.7 x 2.8 cm)

Specifications

For Use With (Equipment)
Universal
Length (Metric)
1.6 cm
Width (Metric)
1.5 cm
Dimensions (LxW)
16 x 15 mm
Product Line
Gene Frame
Type
Microarray Slide Frame
Quantity
100 Coverslips
Thickness (Metric)
0.25 mm
Volume (Metric)
0.65 mL

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text