PCR Plate, 96-well, non-skirted, Sustain™ Series
PCR Plate, 96-well, non-skirted, Sustain™ Series
Thermo Scientific™
PCR Plate, 96-well, non-skirted, Sustain™ Series
Green features
The Thermo Scientific PCR Plate, 96-well, non-skirted, Sustain Series, is for use in PCR and qPCR applications and has aRead more
Have Questions?
Catalog NumberQuantity
AB0600SS25 plates
Catalog number AB0600SS
Price (EUR)
129,00
Each
Add to cart
Quantity:
25 plates
Price (EUR)
129,00
Each
Add to cart
Ask our AI about this Product
The Thermo Scientific PCR Plate, 96-well, non-skirted, Sustain Series, is for use in PCR and qPCR applications and has a 0.3 mL well volume when paired with adhesive films or foils. The plate features a cut corner located at H1 and is compatible with most thermal cyclers.
Features and benefits
  • Non-skirted format compatible with most thermal cyclers
  • Clear well bottom for sample visibility
  • Cut corner: H1
  • 0.3 mL maximum well volume (when used with adhesive or heat seals)
  • Sealable using adhesive films and foils, heat seals, and flat and domed cap strips
A breakthrough in sustainable PCR technology
Thermo Scientific Sustain Series PCR plastics signify a breakthrough in our PCR and quantitative PCR (qPCR) product offering that goes beyond providing accurate and reproducible PCR data. They represent our commitment to integrating more sustainable practices into our manufacturing processes, without compromising on product quality or performance. Our Sustain Series PCR plastics are manufactured under ISO 9001 guidelines within a Class 100,000 cleanroom from a biobased medical-grade virgin polypropylene that is chemically and molecularly identical to our existing Thermo Scientific PCR plastics. These products are supported by certification from the globally recognized International Sustainability and Carbon Certification (ISCC) system. This greener approach is equivalent to 1.96X lower CO2 equivalent emissions per kg of biobased polypropylene resin compared to traditional fossil fuel-based polypropylene resin.
Additional product details
  • Uniform, ultra-thin walls enable maximum and consistent heat transfer for equally high performance of every sample
  • Each well is visually inspected and electrostatically tested for defects and contaminants, and every lot is run through a PCR cycling test to evaluate sealing
  • Manufactured from second generation bio-circular feedstocks that are chemically and molecularly identical to our existing Thermo Scientific PCR plastics
Specifications
AutoclavableAutoclavable
Certifications/ComplianceCertified DNase-, RNase-, Human DNA-, and PCR Inhibitor-free
Green FeaturesLess waste, renewable material
Quantity25 plates
BarcodeCustom Barcoding Available
ColorNatural
FormatSBS Standard
MaterialPolypropylene
No. of Wells96
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.
Documents & Downloads
Certificates
Share catalog number, name or link
1x1 image pixel for data collection