Thermo Scientific™
Domed PCR Caps, strips of 8, natural
Catalog number: AB0602
Have Questions?
Thermo Scientific™
Domed PCR Caps, strips of 8, natural
Catalog number: AB0602
Have Questions?
Catalog Number
AB0602
Unit Size
300 stirps
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
AB0602300 stirps
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Domed PCR Cap, Strips of 8, Natural are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Features of Thermo Scientific Domed PCR Cap Strips
• Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
• Large end tabs for strip removal (12 strips only)
• 25 packs of 12
Related Products
Domed PCR Caps, strips of 8
Domed PCR Caps, strips of 8, blue
Domed PCR Caps, strips of 8, green
Domed PCR Caps, strips of 8, purple
Domed PCR Caps, strips of 8, red
Domed PCR Caps, strips of 8, yellow
Domed PCR Caps, strips of 12
Specifications
Color
Clear
Material
Polypropylene
No. of Wells
8
Quantity
25 packs of 12
Format
Strips of 8 Domed Caps
Product Type
PCR Plate Cap Strip
Certified RNase/DNase-free
Yes
Plate Compatibility
96-well Plate