Thermo Scientific 96-well Low Profile PCR Plate, black, 25 plates, Skirted—The Thermo Scientific 96-Well Full-Skirted Plates have a low-profile, thin well wall design and are compatible with PCR applications. They feature an ANSI:SBS footprint, and the plates are stackable for use in automated systems. The 96-Well Full-Skirted Plates feature a cut corner at the H1 location. These plates also have eight holes in the skirt, aiding in plate positioning and removal from the thermal cycler block. The full skirt surround provides an even surface for robotic arms to grip, making the plate suitable for use with automated systems.
Features of the Thermo Scientific 96-well Low Profile PCR Plate:
• ANSI:SBS footprint and stackable for use in automated systems • Low profile to reduce dead space and increase PCR efficiency • 0.2 mL maximum well volume (when used with adhesive and heat seals) • Sealable using Thermo-Mats, Adhesive Films and Foils, Heat Seals, and all Thermo Scientific Cap Strips • 8 holes in the skirt aid plate positioning and removal from the thermal cycler block • Raised rim design around each well enables secure sealing and safeguards against evaporation • Uniform, thin well walls deliver maximum and consistent heat transfer • Black lettered versions feature black alpha-numeric lettering to allow for easy well identification • Cut Corner: H1