Thermo Scientific™

Flat PCR Caps, strips of 12

Catalog number: AB0851
Have Questions?
Thermo Scientific™

Flat PCR Caps, strips of 12

Catalog number: AB0851
Have Questions?
Catalog Number
AB0851
Unit Size
200 strips
Price (USD)
Quantity
Catalog Number
Unit Size
80 strips
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
AB0851200 strips
AB085080 strips
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Flat PCR Cap ,Strip of 12 are used for PCR applications and work with the Thermo Scientific 96-well PCR plates and strip tubes. These cap strips are available in either 8- or 12-cap strips. The Flat PCR Cap Strips signature flat cap allows you to label each sample.

Features of the Thermo Scientific Flat PCR Cap Strip

• 8- and 12-cap strips
• Suitable for use with 96-well PCR plates as well as standard and low profile strips
• Flat surface of each cap facilitates sample labeling
• 25 packs of 8

Related Products
Flat PCR Caps, strips of 8
Flat PCR Caps, strips of 12
Low Profile Tubes, strips of 12
Tubes, strips of 12

Specifications

Color
Natural
Certifications/Compliance
Certified DNA/RNase Free
Material
Polypropylene
No. of Wells
12
Quantity
25 packs of 8
For Use With (Application)
Thermal Cyclers
Format
200 Strip Caps
For Use With (Equipment)
Thermal Cyclers
Product Type
PCR Plate Cap Strip
Certified RNase/DNase-free
Yes
Plate Compatibility
96-well Plate

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text