Thermo Scientific™
Domed PCR Caps, strips of 12
Catalog number: AB0852
Have Questions?
Thermo Scientific™
Domed PCR Caps, strips of 12
Catalog number: AB0852
Have Questions?
Catalog Number
AB0852
Unit Size
80 strips
Price (USD)
Quantity
Catalog Number
Unit Size
200 strips
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
AB085280 strips
AB0853200 strips
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Scientific Domed PCR Cap, Strips of 12 are compatible with 96-well PCR plates and standard and low profile strip tubes. The 12 strips have large end tabs for easy strip removal.
Features of Thermo Scientific Domed PCR Cap Strips
• Suitable for use with all 96-well PCR plates as well as standard and low profile strip tubes
• Large end tabs for strip removal (12 strips only)
• 10 packs of 8
Related Products
Domed PCR Caps, strips of 8
Domed PCR Caps, strips of 8, blue
Domed PCR Caps, strips of 8, green
Domed PCR Caps, strips of 8, purple
Domed PCR Caps, strips of 8, red
Domed PCR Caps, strips of 8, yellow
Domed PCR Caps, strips of 8, natural
Domed PCR Caps, strips of 12
Low Profile Tubes, strips of 12
Tubes, strips of 12
Specifications
Color
Clear
Material
Polypropylene
No. of Wells
12
Quantity
10 packs of 8
Format
Strips of 12 Domed Caps
Product Type
PCR Plate Cap Strip
Certified RNase/DNase-free
Yes
Plate Compatibility
96-well Plate