Tubes, strips of 12
Tubes, strips of 12
Thermo Scientific™

Tubes, strips of 12

The Thermo Scientific 0.2 mL Strip Tubes are designed for PCR and qPCR applications, and are compatible with 0.2 mLRead more
Have Questions?
Catalog NumberQuantity
AB111210 Packs of 8 Strips
Catalog number AB1112
Price (EUR)
191,00
Each
Add to cart
Quantity:
10 Packs of 8 Strips
Price (EUR)
191,00
Each
Add to cart
Ask our AI about this Product
The Thermo Scientific 0.2 mL Strip Tubes are designed for PCR and qPCR applications, and are compatible with 0.2 mL thermal cycler blocks. While these tubes are sold without caps, Thermo Scientific offers multiple cap options, including an ultra clear cap that's ideal for use in qPCR assays. The caps form a secure seal, yet are easy to apply and remove. Caps increase the overall tube capacity to 0.25 mL

Features of The Thermo Scientific 0.2 mL Strip Tubes

• Compatible with 0.2 mL thermal cycler blocks
• 12 tube strips
• Sealable using flat and domed cap strips. Ultra-Clear cap options available for use in qPCR assays
• 10 packs of 8

Applications

• PCR
• qPCR/Real-Time PCR

Related Products
Tubes, strips of 8
Tubes, strips of 8, white
Tubes, strips of 8, blue
Tubes, strips of 8, green
Tubes, strips of 8, purple
Tubes, strips of 8, red
Tubes, strips of 8, yellow
Flat PCR Caps, strips of 12
Domed PCR Caps, strips of 12
Specifications
Certifications/ComplianceCertified DNA/RNase Free
For Use With (Equipment)Thermal Cyclers
Format80 Strip Tubes
Quantity10 Packs of 8 Strips
Reaction SpeedStandard
Volume (Metric) Working0.2 mL
Closure TypeNot Included
ColorNatural
MaterialPolypropylene
Product Type12-Strip Tube
SterilityNon-sterile
Volume (Metric)0.25 mL
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection