Thermo Scientific™

ABsolute QPCR Mix, no ROX

Catalog number: AB1132B
Have Questions?
Thermo Scientific™

ABsolute QPCR Mix, no ROX

Catalog number: AB1132B
Have Questions?
Catalog Number
AB1132B
also known as AB-1132/B
Unit Size
1,600 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
400 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AB1132B
also known as AB-1132/B
1,600 reactions
-
AB1133A400 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
**********************
Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of ABsolute QPCR Mix and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific ABsolute qPCR Mixes are optimized for use with hybridization probes and contain all the components necessary to perform quantitative PCR, with the exception of template and primers/probes. The 2X master mix contains a proprietary reaction buffer that provides high sensitivity, specificity, and reproducibility. ABsolute qPCR Mixes are available with the option of included high ROX reference dye concentration, Low ROX concentration, and with no ROX reference dye. There is also a master mix version optimized for capillary instruments.

Highlights

• High sensitivity & specificity - The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase
• Reproducibility - All mixes undergo extensive QC to ensure batch to batch consistency
• Optimised buffer - For maximal performance in multiplex reactions

Applications
• Gene expression analysis
• siRNA validation
• Genotyping

Related Products
ABsolute QPCR Mix, no ROX
ABsolute QPCR Mix, low ROX
ABsolute QPCR Mix, ROX
ABsolute QPCR Capillary Mix
ABsolute QPCR Mix, SYBR Green, no ROX
ABsolute QPCR Mix, SYBR Green, low ROX
ABsolute QPCR Mix, SYBR Green, ROX
ABsolute QPCR Mix, SYBR Green, fluorescein
ABsolute QPCR Capillary Mix, SYBR Green
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Description
ABsolute QPCR Mix, QPCR mix; 1.25mL vial; 800 rxns
Detection Method
Primer-probe
GC-Rich PCR Performance
High
PCR Method
qPCR
Polymerase
Taq DNA Polymerase
Reaction Speed
Standard
For Use With (Equipment)
BioRad CFX96™, BioRad iCycler iQ, BioRad iQ5, BioRad MyiQ, MJ Chromo4, Eppendorf MasterCycler RealPlex, Corbett RotorGene, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, MJ Opticon
Product Line
ABsolute
Product Type
qPCR Mix
Quantity
16 x 1.25 mL vials; 800 x 50 μL reactions
Sufficient For
1600 Reactions
For Use With (Application)
Gene Expression, Genotyping, siRNA Validation
Concentration
2X
Fidelity (vs. Taq)
2 X
No. of Reactions
1600 Reactions
Sample Type
DNA, dsDNA
Volume
1.25 mL

Contents & Storage

Contains:
• 16 x 1.25 mL ABsolute QPCR Mix (2X)

Store at -20°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text