Thermo Scientific™

ABsolute QPCR Mix, SYBR Green, no ROX

Catalog number: AB1158B
Have Questions?
Thermo Scientific™

ABsolute QPCR Mix, SYBR Green, no ROX

Catalog number: AB1158B
Have Questions?
Catalog Number
AB1158B
also known as AB-1158/B
Unit Size
1,600 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
AB1159A
also known as AB-1159/A
Unit Size
400 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AB1158B
also known as AB-1158/B
1,600 reactions
-
AB1159A
also known as AB-1159/A
400 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
**********************
Alternative Product: Try PowerUp SYBR Green Master Mix, our newest, high-performance, SYBR dye-based master mix for superior performance at a very competitive price. With PowerUp SYBR Green Master Mix, we’ve taken the best of ABsolute QPCR Mix, SYBR Green, and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific ABsolute qPCR SYBR Green Mixes are optimized for SYBR Green chemistry and contain all the components necessary to perform quantitative PCR, with the exception of template and primers. The 2X qPCR SYBR Green Mix contains optimal levels of active SYBR Green I dye and Thermo-Start DNA Polymerase supplied in a proprietary reaction buffer that enables detection of low copy number targets.

ABsolute qPCR SYBR Green Mixes are available with the option of included high ROX reference dye concentration, Low ROX concentration, Fluorescein or no reference dye. A master mix optimized for capillary instruments is also available.

Highlights

• High Sensitivity & Specificity—The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase
• Optimised buffer containing SYBR Green I
• Reproducibility—All master mixes undergo extensive QC to ensure batch to batch consistency
• Broad dynamic range

Applications

• Gene expression analysis
• siRNA validation
• Genotyping

Related Products
ABsolute QPCR Mix, no ROX
ABsolute QPCR Mix, low ROX
ABsolute QPCR Mix, ROX
ABsolute QPCR Capillary Mix
ABsolute QPCR Mix, SYBR Green, no ROX
ABsolute QPCR Mix, SYBR Green, low ROX
ABsolute QPCR Mix, SYBR Green, ROX
ABsolute QPCR Mix, SYBR Green, fluorescein
ABsolute QPCR Capillary Mix, SYBR Green
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
SYBR
GC-Rich PCR Performance
High
PCR Method
qPCR
Polymerase
Taq DNA Polymerase
Reaction Speed
Standard
For Use With (Equipment)
BioRad CFX96™, MJ Chromo4, MJ Opticon, Eppendorf MasterCycler RealPlex, Bio-Rad CFX384 Touch™, Roche LightCycler 480, Cepheid SmartCycler, Corbett RotorGene
Product Line
ABsolute
Product Type
Real Time PCR SYBR Master Mix
Sufficient For
1600 Reactions of 25 μL Each
For Use With (Application)
Gene Expression
Compatibility
qPCR Cyclers
Concentration
2X
No. of Reactions
1,600 Reactions
Sample Type
DNA, dsDNA
Volume
16 x 1.25 mL

Contents & Storage

Contains:
• 16 x 1.25 mL ABsolute QPCR SYBR Green Mix (2X)
• 100 μL MgCl2 (1M)
Sufficient for 400 x 25 μL reactions.

Store at -20°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text