You need to enable JavaScript to run this app.
Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
Home
Shop All Products
Microplates
Microplate Covers
Microplate Well Caps
ABgene Individual Storage Plate Caps
Thermo Scientific™
ABgene Individual Storage Plate Caps
Catalog number:
AB1179
Related applications:
Cell Counting, Viability, & Cryopreservation
Microplates, Dishes & Flasks
Have Questions?
Contact Us
Thermo Scientific™
ABgene Individual Storage Plate Caps
Catalog number:
AB1179
Related applications:
Cell Counting, Viability, & Cryopreservation
Microplates, Dishes & Flasks
Have Questions?
Contact Us
Catalog Number
AB1179
also known as AB-1179
Unit Size
Case of 960
Price (USD)
Request A Quote
Save to list
Catalog Number
Unit Size
Price (USD)
AB1179
also known as AB-1179
Case of 960
Request A Quote
Save to list
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
Thermo Fisher Individual Storage Plate Caps are designed for sealing storage plates and cluster tubes.
Features of Thermo Fisher Individual Storage Plate Caps
• Low profile for increased well volume
• End tabs for ease of removal
• Individual Caps
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Color
Natural
Material
Polypropylene
Sterility
Cleanroom
Autoclavable
Autoclavable
Product Type
Plate Lid
Product Line
Abgene
Certified RNase/DNase-free
Yes
Pierceable
No
Well Shape
Round
Plate Compatibility
Abgene 96 Round Well Plate
Figures
Documents & Downloads
Certificates
Search by lot number or partial lot number
Search
Request a Certificate
Frequently asked questions (FAQs)
Citations & References
Search citations by name, author, journal title or abstract text
Search