**********************Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’veRead more
Have Questions?
Change view
Catalog Number
No. of Reactions
AB1319A
400 Reactions
AB1318B
1600 Reactions
2 Options
Catalog number AB1319A
Price (USD)
918.00
Each
Add to cart
No. of Reactions:
400 Reactions
Price (USD)
918.00
Each
Add to cart
********************** Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of ABsolute QPCR Mix and added additional capabilities for your gene expression analysis.
********************** Thermo Scientific ABsolute qPCR Mixes are optimized for use with hybridization probes and contain all the components necessary to perform quantitative PCR, with the exception of template and primers/probes. The 2X master mix contains a proprietary reaction buffer that provides high sensitivity, specificity, and reproducibility. ABsolute qPCR Mixes are available with the option of included high ROX reference dye concentration, Low ROX concentration, and with no ROX reference dye. There is also a master mix version optimized for capillary instruments.
Highlights
• High sensitivity & specificity - The master mixes contain our patented hot start enzyme Thermo-Start DNA Polymerase • Reproducibility - All mixes undergo extensive QC to ensure batch to batch consistency • Optimised buffer - For maximal performance in multiplex reactions