Thermo Scientific™

PCR Plate, 384-well, standard, blue

Catalog number: AB1384B
Have Questions?
Thermo Scientific™

PCR Plate, 384-well, standard, blue

Catalog number: AB1384B
Have Questions?
Catalog Number
AB1384B
Unit Size
50 plates
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
AB1384B50 plates
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Standard Thermo Scientific 384-well plate, blue, is compatible with the majority of 384 block PCR and sequencing instruments. It has a maximum well volume of 40 μL and a working volume of 25 μL. This 384-well plate is compatible with all major 384 block thermal cyclers, and it has a cut corner at the A24 location.

Highlights of the Standard Thermo Scientific 384-well plate

• Fully skirted for use with automated systems
• Blue color to aid in sample tracking
• Compatible with all leading 384 block thermal cyclers
• 40 μL maximum well volume, 25 μL working volume
• Raised rim design around each well enables secure sealing and safeguards against evaporation
• Uniform, thin well walls deliver maximum and consistent heat transfer
• White plates are optimized for qPCR by increasing sensitivity and reducing variability in fluorescence detection
• Cut corner: A24

Applications

• PCR
• qPCR/Real-Time PCR

Specifications

Barcode
No Barcode
Certifications/Compliance
Certified RNase/DNase-free
Packaging
50 Plate
Color
Blue
Product Type
PCR Microplate
For Use With (Equipment)
Real-Time PCR Systems or Thermal Cyclers
Skirt Style
Skirted
Material
Polypropylene
Volume (Metric) Well
40 μL
No. of Wells
384
Volume (Metric) Working
25 μL
Reaction Speed
Standard

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text