Thermo Scientific™

VersiPlate Frame, 96-well, skirted, white

Catalog number: AB1805
Have Questions?
Thermo Scientific™

VersiPlate Frame, 96-well, skirted, white

Catalog number: AB1805
Have Questions?
Catalog Number
AB1805
Unit Size
25 plates
Price (USD)
Quantity
Catalog NumberUnit SizePrice (USD)Quantity
AB180525 plates
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific VersiPlate Frame is a reusable polycarbonate frame that serves as a useful rack for a VersiPlate or any number of low profile strip tubes facilitating sample loading into thermal cyclers.

Highlights

• Easier sample handling as multiple strips of samples can be handled simultaneously
• Reusable polycarbonate frame is suitable for a VersiPlate or other low profile strip tubes
• Can be used with all Thermo Scientific VersiCap™ Mats, cap strips, adhesive and heat seals

Applications

• PCR
• qPCR/Real-Time PCR

Related Products
VersiPlate PCR Strip Tube Plate, 96-well, low profile
VersiPlate PCR Strip Tube Plate, 96-well, low profile, white

Specifications

Barcode
No Barcode
Packaging
25 Plate
Product Line
VersiPlate
Color
White
Product Type
Frame
For Use With (Equipment)
Real-Time PCR Systems or Thermal Cyclers
Material
Polycarbonate
No. of Wells
96
Plate Compatibility
96-Well Plates
Reaction Speed
Low profile

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text