Thermo Scientific™

Verso 1-step RT-qPCR Kit, SYBR Green, ROX

Catalog number: AB4105C
Have Questions?
Thermo Scientific™

Verso 1-step RT-qPCR Kit, SYBR Green, ROX

Catalog number: AB4105C
Have Questions?
Catalog Number
AB4105C
Unit Size
400 reactions
Price (USD)
Availability
-
Quantity
Catalog Number
Unit Size
200 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AB4105C400 reactions
-
AB4105A200 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Thermo Scientific Verso 1-step RT-qPCR ROX Kit for SYBR Green chemistry offers a quick and simple way to detect mRNA. The Verso RT enzyme delivers high processivity and can be used at temperatures up to 57°C, delivering effective transcription through difficult RNA secondary structures.

Features of the Verso 1-step RT-qPCR SYBR Green ROX Kit:

• Eliminate the need for DNase treatments—RT Enhancer (optional) prevents gDNA carry-over
• RT and qPCR steps performed in single tube—convenience and reduced contamination risk
• Use of the entire RT reaction as template for qPCR—improved sensitivity
• Verso RT enzyme is highly sensitive with broad dynamic range—accurate detection of RNA from 1 pg-1 ug
• Verso RT enzyme has a wide working temperature range (42 to 57°C)—improved success with GC-rich and other difficult templates
• Contains an inert blue dye for ease of pipetting during reaction setup

An optional RT Enhancer is also provided and can be used during the RT step to remove any DNA contamination, eliminating the need for DNase I treatment. All Verso 1-step RT-qPCR kits incorporate an inert dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof.

In a 1-step approach to RT-qPCR, both the RT step and qPCR steps are performed in a single tube (1-step RT-qPCR). Adopting a 1-step approach is ideal for primer sequence optimization and collecting comparative data in experiments that do not require subsequent cDNA storage. The approach minimizes the risk of contamination, and improvements in reproducibility may be observed as a result of reduced sample handling.

Verso 1-Step RT-qPCR Kits for SYBR Green chemistry are compatible with all major real-time thermal cyclers.

Applications

• Gene expression analysis
• RNAi validation
• Microarray validation
• Pathogen detection
• Genetic testing
• Disease research

Related Products
Verso 1-step RT-qPCR Kit, SYBR Green, low ROX
Verso 1-step RT-qPCR Kit, SYBR Green, ROX
Verso 1-step RT-qPCR Kit, SYBR Green, with separate ROX vial
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
SYBR
Format
Tube
GC-Rich PCR Performance
High
PCR Method
1-step RT-qPCR
For Use With (Equipment)
7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
Product Line
Verso
Product Type
1-Step RT-qPCR Kit
Sufficient For
400 Reactions
For Use With (Application)
Gene Expression, Pathogen Detection
No. of Reactions
400 Reactions
Sample Type
RNA

Contents & Storage

Contains:
•1 x 100 μL Verso Enzyme Mix
•1 x 500 μL RT enhancer
•1 x 5 mL 2X 1-Step qPCR SYBR ROX Mix
• 1 x 100μL 1M MgCl2.
Sufficient for 400 x 25 μL reactions

Store at -20°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text