Verso 1-step RT-qPCR Kit, SYBR Green, low ROX
Product Image
Thermo Scientific™

Verso 1-step RT-qPCR Kit, SYBR Green, low ROX

The Thermo Scientific Verso 1-step RT-qPCR Low ROX Kit for SYBR Green chemistry offers a quick and simple way toRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberNo. of Reactions
AB4106C
also known as AB-4106/C
384 Reactions
AB4106A
also known as AB-4106/A
200 Reactions
Catalog number AB4106C
also known as AB-4106/C
Price (USD)
1,432.00
Each
Add to cart
No. of Reactions:
384 Reactions
Price (USD)
1,432.00
Each
Add to cart
The Thermo Scientific Verso 1-step RT-qPCR Low ROX Kit for SYBR Green chemistry offers a quick and simple way to detect mRNA. The Verso RT enzyme delivers high processivity and can be used at temperatures up to 57°C, delivering effective transcription through difficult RNA secondary structures.

Features of the Verso 1-step RT-qPCR SYBR Green Low ROX Kit:

• Eliminate the need for DNase treatments—RT Enhancer (optional) prevents gDNA carry-over
• RT and qPCR steps performed in single tube—convenience and reduced contamination risk
• Use of the entire RT reaction as template for qPCR—improved sensitivity
• Verso RT enzyme is highly sensitive with broad dynamic range—accurate detection of RNA from 1 pg-1 ug
• Verso RT enzyme has a wide working temperature range (42 to 57°C)—improved success with GC-rich and other difficult templates
• Contains an inert blue dye for ease of pipetting during reaction setup

An optional RT Enhancer is also provided and can be used during the RT step to remove any DNA contamination, eliminating the need for DNase I treatment. All Verso 1-step RT-qPCR kits incorporate an inert dye to significantly enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof.

In a 1-step approach to RT-qPCR, both the RT step and qPCR steps are performed in a single tube (1-step RT-qPCR). Adopting a 1-step approach is ideal for primer sequence optimization and collecting comparative data in experiments that do not require subsequent cDNA storage. The approach minimizes the risk of contamination, and improvements in reproducibility may be observed as a result of reduced sample handling.

Verso 1-Step RT-qPCR Kits for SYBR Green chemistry are compatible with all major real-time thermal cyclers.

Applications

• Gene expression analysis
• RNAi validation
• Microarray validation
• Pathogen detection
• Genetic testing
• Disease research

Related Products
Verso 1-step RT-qPCR Kit, SYBR Green, low ROX
Verso 1-step RT-qPCR Kit, SYBR Green, ROX
Verso 1-step RT-qPCR Kit, SYBR Green, with separate ROX vial
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)7500 System, QuantStudio™, ViiA™ 7 System, Stratagene Mx4000, Stratagene Mx3000P, Stratagene Mx3005P
FormatTube
No. of Reactions384 Reactions
Product LineVerso
Product TypeReal Time PCR Kit
Quantity400 reactions
Sample TypeRNA
Volume100 μL
Detection MethodSYBR
GC-Rich PCR PerformanceHigh
PCR Method1-step RT-qPCR
Unit SizeEach
Contents & Storage
Contains:
• 1 x 100 μL Verso Enzyme Mix
• 1 x 500 μL RT enhancer
• 1 x 5 mL 2X 1-Step qPCR SYBR Low ROX Mix

Store at -20°C

Share catalog number, name or link