Thermo Scientific™

ABsolute Blue QPCR Mix, ROX

Catalog number: AB4139A
Have Questions?
Thermo Scientific™

ABsolute Blue QPCR Mix, ROX

Catalog number: AB4139A
Have Questions?
Catalog Number
AB4139A
also known as AB-4139/A
Unit Size
Price (USD)
Availability
-
Quantity
Catalog Number
AB4138B
also known as AB-4138/B
Unit Size
1,600 reactions
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AB4139A
also known as AB-4139/A
-
AB4138B
also known as AB-4138/B
1,600 reactions
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
**********************
Alternative Product: Try TaqMan Fast Advanced Master Mix, our highest-performance, probe-based master mix. With TaqMan Fast Advanced Master Mix, we’ve taken the best of ABsolute Blue QPCR Mix and added additional capabilities for your gene expression analysis.

**********************
Thermo Scientific ABsolute Blue qPCR Mixes are 2X master mixes for probe chemistry. There are suitable alternatives available for all common real-time instruments. ABsolute Blue qPCR Mixes utilize Thermo-Start Taq hot start enzyme for increased sensitivity. The master mixes incorporate an inert blue dye to enhance the contrast between reagent and plastic, making verification of master mix dispensing quick, easy, and foolproof.

Highlights

High performance—ABsolute sensitivity, consistency & convenience
Enhanced contrast and reduced errors—easily identify correctly aliquotted reaction mix in all plates and tubes
Universal—compatible with all major qPCR platforms
Adaptable—available with different ROX passive reference dye concentrations

Applications

• Gene expression analysis
• siRNA validation
• Genotyping

Related Products
ABsolute Blue QPCR Mix, low ROX
ABsolute Blue QPCR Mix, with separate ROX vial
ABsolute Blue QPCR Mix, ROX
ABsolute Blue QPCR Mix, SYBR Green, low ROX
ABsolute Blue QPCR Mix, SYBR Green, ROX
ABsolute Blue QPCR Mix, SYBR Green, with separate ROX vial
ABsolute Blue QPCR Mix, SYBR Green, fluorescein
For Research Use Only. Not for use in diagnostic procedures.

Specifications

Detection Method
Primer-probe
Format
Tube
GC-Rich PCR Performance
High
PCR Method
qPCR
Polymerase
Taq DNA Polymerase
Reaction Speed
Standard
For Use With (Equipment)
7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
Product Line
ABsolute Blue
Product Type
qPCR Mix
Sufficient For
400 Reactions
Concentration
2X
Fidelity (vs. Taq)
2 X
No. of Reactions
400 Reactions
Sample Type
DNA, dsDNA
Volume
5 mL

Contents & Storage

Contains:
• 1 x 5 mL ABsolute Blue qPCR ROX Mix (2X)
Sufficient for 400 x 25 μL reactions.

Store at -20°C

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text