Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Measure common colorimetric water and wastewater parameters using the Thermo Scientific™ Orion™ AQUAfast AQ4000 Colorimeter. This instrument has 65 pre-programmed methods, automatic selection of four wavelengths and timer. Use auto-test ampoules with Auto-ID barcodes to simplify sample preparation and tests. Ampoules contain pre-measured liquid reagent and fill with the exact volume of sample. The colorimeter reads the barcode and automatically identifies the parameter and initiates the test. This IP67 waterproof meter records 100 data points and offers an average battery life of 2,500 hours intermittent use.
Description | 16mm vials, pk. of 10 |
Type | Vials |
For Use With (Equipment) | AQ4000 Colorimeter |
Length (English) | 0.6 in. |
Length (Metric) | 16 mm |
Quantity | 10/Pk. |
Unit Size | Case of 10 |
Description | 13mm Vial Adapter for AQUAfast AQ4000 Colorimeter |
For Use With (Equipment) | AQ4000 Colorimeter |
Range | 01 to 0.25 mg/L |
Unit Size | Each |
Description | 16mm Vial Adapter for AQUAfast AQ4000 Colorimeter |
For Use With (Equipment) | AQ4000 Colorimeter |
Unit Size | Each |
Absorbance Range | 0 to 2 A |
Battery Life | 2500 hr (Alkaline); 10000 hr (Lithium) |
Cuvette Details | 13, 16, 24 mm |
Description | Colorimeter for use with 189 pre-programmed method |
Battery Type | AA |
Display | Custom LCD |
IP Rating | IP67 |
Light Source | LED |
Wavelength Range | 420, 520, 580, 610 nm |
Wavelength Selection | Automatic |
Humidity | 90 at 50°C max. % |
Includes | AQ4000 Colorimeter, 13 and 16 mm vial adapter, tamping rod, 16 mm vial, zero kit (AQ4ZER), 4 AA Batteries and carrying case |
Keypad | Keypad 12 dual function keys with tactile feedback |
Outputs | 100 |
Resolution | ±2 nm |
Temperature Range (Metric) | 0°C to 45°C |
Test Method | Up to 190 and 10 user defined programmed methods |
Unit Size | Each |
Description | Zero kit |
Type | Zero Kit |
For Use With (Equipment) | AQ4000 Colorimeter |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Description | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
AC2V16 | Case of 10 | 16mm vials, pk. of 10 | 96.30 Online price 107.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AC4A13 | Each | 13mm Vial Adapter for AQUAfast AQ4000 Colorimeter | 169.20 Online price 188.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AC4A16 | Each | 16mm Vial Adapter for AQUAfast AQ4000 Colorimeter | 169.20 Online price 188.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AQ4000 | Each | Colorimeter for use with 189 pre-programmed method | 1,476.00 Online price 1,640.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AQ4ZER | Each | Zero kit | 69.12 Online price 76.80 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
Features:
Benefits:
Ordering Information:
For use with Orion™ AQUAfast™ AQ4000, Orion™ AQUAfast™ AQ3070, Orion™ AQUAfast™ AQ3700 and discontinued Orion AQ2 series meters.
Note: Uses Thermo Scientific™ Auto-ID™ ampoules, liquid, powder and tablet reagent including many methods accepted for EPA, ISO and other regulatory reporting.