Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Type | Includes |
---|---|---|
AC3019 | Potassium | |
AC2002 | Alkalinity | 100 Alkalinity-M Tablets |
AC3002P | CaCO3 | |
AC2027 | Aluminum | Set of 2: 50 Aluminum No 1 Tablets, 50 Aluminum No |
AC4P27 | Aluminum | |
AC2012 | Ammonia | Set of 2: 50 Ammonia No 1 Tablets, 50 Ammonia No 2 |
AC2035 | Reagent Tablet | Bromine |
AC2017 | Chloride | Set of 2: 50 Chloride T1 Tablets, 50 Chloride T2 T |
AC2099 | Chlorine Dioxide | 100 DPD No 1 Tablets |
AC2071 | Chlorine | 100 DPD No 1 Tablets |
AC2070 | Chlorine | Set of 2: 50 DPD No 1 Tablets, 50 DPD No 3 Tablets |
AC2072 | Chlorine | 100 DPD No 4 Tablets |
AC3072 | Chlorine, Sodium Chlorate | |
AC2029 | Copper | Set of 2: 50 Copper No 1 Tablets, 50 Copper No 2 Tablets |
AC2065 | Zinc | Set of 2: 50 Copper/Zinc LR Tablets, 50 EDTA Table |
AC2098 | Cyanuric Acid | |
AC3032C2 | Calcium | |
AC3032C | Calcium | |
AC3032T | Metallphthalein Method | |
AC2030 | Hydrazine | Kit: Hydrazine Powder, Spoon |
AC3069 | Hydrogen Peroxide | |
AC2078 | Iron | 100 Iron LR Tablets |
AC2055 | Manganese | Set of 2: 50 Manganese LR1 Tablets, 50 Manganese L |
AC2007 | Nitrate | 50 x Nitrite HR No. 1 Tablets, 50 x Nitrite HR No. 2 Tablets |
AC2046 | Nitrite | 100 Nitrite LR Tablets |
AC2048 | Ozone | 100 Indigo Blue Tablets |
AC2001 | pH | 100 Phenol Red Tablets |
AC3001BP | Bromocresol Purple, pH, Low Range | |
AC3001TB | Thymol Blue, pH, High Range | |
AC2095-WA | Phosphate | Set of 2: 50 Phosphate No 1 LR Tablets, 50 Phosphate No 2 LR Tablet |
AC2096 | Phosphate | Set of 2: 50 Phosphate No 1 HR Tablets, 50 Phosphate No 2 HR Tablets |
AC2060 | Silica | Set of 2: 50 Silica No 1 Tablets, 50 Silica No 2 T |
AC2061 | Silica | 100 Silica Phosphate Removal Tablets |
AC2082 | Sulfate | 100 Sulfate Tablets |
AC2016 | Sulfide | Set of 2: 100 Sulfide No 1 Tablets, 100 Sulfide No 2 |
Choose the reagent tablets you need for Orion™ AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers with Thermo Scientific™ Orion™ AQUAfast II Chemistries Test Kits. These convenient, rapidly dissolving tablets can be precisely added to samples without leaving residual reagent in the packaging and are very stable for long shelf life accessibility. Tablet reagents measure alkalinity, aluminum, ammonia, boron, chloride, chlorine, chlorine dioxide, copper, DEHA, hardness, hydrazine, hydrogen peroxide, iron, manganese, nitrate, nitrite, ozone, pH, phosphate, potassium, silica, sulfate, sulfide and zinc.
Share catalog number, name or link