Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Catalog Number | Type | Range |
---|---|---|
AC4P12 | Ammonia | 0.01 to 0.8 mg/L N |
AC4P29 | Copper (free) | 0.05 to 5 mg/L Cu |
AC4P42 | Molybdate/Molybdenum | 0.5 to 66 mg/L MoO4 |
AC4P46 | Nitrite | 0.01 to 0.3 mg/L N |
AC4P55 | Manganese | 0.1 to 18 mg/L Mn |
AC4P60 | Silica | 1 to 90 mg/L SiO2 |
AC4P71 | Chlorine (free) | 0.02 to 2 mg/L Cl2 |
AC4P72 | Chlorine (total) | 0.02 to 2 mg/L Cl2 |
AC4P78 | Iron (ferro) | 0.02 to 3 mg/L Fe |
AC4P79 | Iron (total) | 0.02 to 1.8 mg/L Fe |
AC4P82 | Sulfate | 5 to 100 mg/L SO4 |
AC4P95 | Phosphate (ortho) | 0.06 to 2.5 mg/L PO4 |
Use Thermo Scientific™ Orion™ AQUAfast Powder Pack Reagents with Orion AQUAfast colorimeters and Thermo Scientific Orion AquaMate Vis and UV-Vis spectrophotometers. Powder pack chemistries dissolve quickly in solution, are easy to mix and do not require a stir rod or crushing. Each powder pack is notched for simplified pouch opening in the lab or field. Powder pack reagents measure aluminum, ammonia, chlorine, copper, cyanide, DEHA, iron, manganese, molybdate, nitrite, phosphate, silica and sulfate.
Share catalog number, name or link