Search Thermo Fisher Scientific
Measure residual chlorine in water samples using Thermo Scientific™ Orion™ AQUAfast Free and Total Chlorine Bulk DPD Powder Dispensers and Vials with Orion AQUAfast colorimeters and Thermo Scientific™ Orion™ AquaMate Vis and UV-Vis spectrophotometers. The powder chemistries dissolve quickly in the solution and are easy to mix. Each vial contains 250 counts of DPD reagent and is available for free chlorine or total chlorine analysis in the lab or field.
Description | AQUAfast total chlorine DPD powder refill for AQ250T dispenser, 2 vials x 250 count each |
For Use With (Equipment) | AQUAfast AQ250T total chlorine DPD bulk powder dispenser |
Includes | Two total chlorine powder vials, 500 count (2 x 250 count) |
Range | 0.02 to 2 mg/L Cl2 |
Reagent Form | Powder |
Reagents | Chlorine |
Test Method | DPD |
Unit Size | Each |
Description | AQUAfast free chlorine DPD bulk powder dispenser with one vial x 250 count |
For Use With (Equipment) | Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ4000 colorimeter, Orion AQUAfast AQ3700 colorimeter, Orion AQUAfast AQ3170 colorimeter, Orion AQUAfast AQ3070 (obsolete) |
Includes | DPD bulk powder dispenser and one free chlorine powder vial, 250 count |
Range | 0.02 to 2 mg/L Cl2 |
Reagent Form | Powder |
Reagents | Chlorine |
Test Method | DPD |
Unit Size | Each |
Description | AQUAfast free chlorine DPD powder refill for AQ250F dispenser, 1 vial x 250 count |
For Use With (Equipment) | AQUAfast AQ250F free chlorine DPD bulk powder dispenser |
Includes | One free chlorine powder vial, 250 count |
Range | 0.02 to 2 mg/L Cl2 |
Reagent Form | Powder |
Reagents | Chlorine |
Test Method | DPD |
Unit Size | Each |
Description | AQUAfast free chlorine DPD powder refill for AQ250F dispenser, 2 vials x 250 count each |
For Use With (Equipment) | AQUAfast AQ250F free chlorine DPD bulk powder dispenser |
Includes | Two free chlorine powder vials, 500 count (2 x 250 count) |
Range | 0.02 to 2 mg/L Cl2 |
Reagent Form | Powder |
Reagents | Chlorine |
Test Method | DPD |
Unit Size | Each |
Description | AQUAfast total chlorine DPD powder dispenser with one vial x 250 count |
For Use With (Equipment) | Orion AquaMate Vis and UV-Vis Spectrophotometers, Orion AQUAfast AQ3700 Colorimeter, Orion AQUAfast AQ3170 Colorimeter |
Includes | DPD bulk powder dispenser and one total chlorine powder vial, 250 count |
Range | 0.02 to 2 mg/L Cl2 |
Reagent Form | Powder |
Reagents | Chlorine |
Test Method | DPD |
Unit Size | Each |
Description | AQUAfast total chlorine DPD powder refill for AQ250T dispenser, 1 vial x 250 count |
For Use With (Equipment) | AQUAfast AQ250T total chlorine DPD bulk powder dispenser |
Includes | One total chlorine powder vial, 250 count |
Range | 0.02 to 2 mg/L Cl2 |
Reagent Form | Powder |
Reagents | Chlorine |
Test Method | DPD |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Includes | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
AC72P2 | Each | Two total chlorine powder vials, 500 count (2 x 250 count) | 108.90 Online price 121.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AQ250F | Each | DPD bulk powder dispenser and one free chlorine powder vial, 250 count | 75.33 Online price 83.70 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AC71P1 | Each | One free chlorine powder vial, 250 count | 57.69 Online price 64.10 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AC71P2 | Each | Two free chlorine powder vials, 500 count (2 x 250 count) | 108.90 Online price 121.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AQ250T | Each | DPD bulk powder dispenser and one total chlorine powder vial, 250 count | 75.33 Online price 83.70 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
AC72P1 | Each | One total chlorine powder vial, 250 count | 57.69 Online price 64.10 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|