Thermo Scientific™
AccuLINK™ Software Package
Catalog number: ACCULINK
Have Questions?
Thermo Scientific™
AccuLINK™ Software Package
Catalog number: ACCULINK
Have Questions?
The Thermo Scientific™ AccuLINK Software Package continuously compares online analytical data with lab values to implement timely automatic calibrations and ensure the highest possible online accuracy in cement analysis. It references neutron activation analysis data measured in real-time with the Thermo Scientific™ CB Omni Agile Online Elemental Analyzer to gold standard X-Ray Fluorescence (XRF) data for cement composition measured with the ARL series of laboratory analyzers (or with instruments from alternative suppliers) to enable consistent operation to an optimal set-point. Use AccuLINK to access lab accuracy online, with minimal manual input, for highly effective diagnostics and troubleshooting and to safeguard operation at a setpoint that maximizes profitability.
Catalog Number
ACCULINK
Unit Size
Each
Description
AccuLINK Software Package
Price (USD)
Full specifications
DescriptionAccuLINK Software Package
Unit SizeEach
Showing 1 of 1
Catalog NumberSpecificationsUnit SizeDescriptionPrice (USD)
ACCULINKFull specifications
EachAccuLINK Software PackageRequest A Quote
DescriptionAccuLINK Software Package
Unit SizeEach
Showing 1 of 1
Reflecting our capabilities as a one-stop shop for laboratory and online elemental analysis for cement, AccuLINK continuously calibrates your online crossbelt analyzer ensuring it consistently operates at peak accuracy, for optimum operational efficiency. Key features include:
  • Easy-to-use, for enhanced data quality with minimal input from lab personnel
  • Flexible, in-depth statistical and graphical data analysis including robust scatter and time series plots, for productive information gathering with minimal training
  • Fully automatic calibration updates, for consistently high online accuracy
  • Automatic outlier screening, for rigorous and robust data analysis