ALPHA-7A Alpha Air Monitors
ALPHA-7A Alpha Air Monitors
Actual product may vary
Thermo Scientific™

ALPHA-7A Alpha Air Monitors

Thermo Scientific™ ALPHA-7A Air Monitor is designed to provide early warning to workers exposed to airborne releases of alpha-emitting radionuclides.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDescription
ALPHA7LAlpha CAM w/Amphenol connector
ALPHA7AAlpha CAM w/o Amphenol connector
ALPHA7OPT2Detector head, Smart remote radial entry
ALPHA7OPT3Detector head (Smart remote in-line)
ALPHA7ADS3Warranty plans; For Alpha-7A or Alpha-7L
Catalog number ALPHA7L
Price (USD)
-
Request A Quote
Description:
Alpha CAM w/Amphenol connector
Ask our AI about this Product

Thermo Scientific™ ALPHA-7A Air Monitor is designed to provide early warning to workers exposed to airborne releases of alpha-emitting radionuclides.

Modern, PC-based continuous air monitor provides faster, more powerful algorithms for the identification and quantification of airborne releases of alpha-emitting radionuclides, primarily transuranics such as 238Pu and 239Pu.

The ALPHA-7A has adjustable alarms, at both DAC and DAC-hour levels, to rapidly warn workers of potentially dangerous releases. In addition to monitoring work areas, it is also an excellent solution for monitoring stacks and ducts.
  • Can serve as a stand-alone CAM or be incorporated into Ethernet-based network
  • Simultaneously monitors up to 8 isotopes
  • Advanced peakshape algorithms; calculates isotopic activity by mapping peaks rather than using regions of interest (ROI)
  • Alpha-spectral data updated every second
  • Concentration, dose, and activity alarms
  • Pentium-class PC-based, Windows 2000 operating system
  • Removable filter holder cartridge
  • High-speed connections via 10-base T Ethernet connection
  • 4 to 20mA analog input and output capacity
  • High-visibility display for status and messages
  • New isotopes easily added from library using any ODBC database manager
  • Automatic gain control based on naturally occurring peaks
  • Tracks alpha-emitting daughter products
  • Full-featured, Windows-based client software presents graphical displays of spectrum and status to any networked or local PC

Two detector designs:

  • Radial entry head for ambient air monitoring

  • Inline head for process or stack monitoring applications

  • Either head may be used remotely from the central display and control unit

Compatible with:

Thermo Scientific RAP-1 pump

Specifications
Certifications/CompliancePassed ANSI N42.17B testing in the US; RadNet compliant. CE qualified for European and other international operations
DescriptionAlpha CAM w/Amphenol connector
Alpha/Beta/Gamma MeasurementAlpha Measurement
For Use With (Application)Alpha Monitoring
Detector TypeAlpha-emitting Radionuclides
Radiation TypeAlpha
Product TypeAlpha Air Monitor
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

0 results displayed, search above for a specific certificate

Share catalog number, name or link

1x1 image pixel for data collection