Vortex Adapter-60

Catalog number: AM10014
Have Questions?

Vortex Adapter-60

Catalog number: AM10014
Have Questions?
Catalog Number
AM10014
Unit Size
1 each
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AM100141 each
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Ambion® Vortex Adapter-60 enables hands-free mixing for up to 60 microfuge tubes or one 96-well plate. The unique shaking motion made possible by this adapter promotes rapid and thorough tissue digestion and so is strongly recommended for use with our MELT™ Total RNA Isolation Kit (SKU #AM1983). The Vortex Adapter-60 consists of a 6 inch (152 mm) Platform and 2 different Microplate Foam Inserts. Each insert fits snugly into the platform to accommodate either 60 microtubes or one standard microplate. The Vortex Adapter-60 is compatible with the Vortex-Genie® 2, Vortex-Genie 2T, and VWR® Mini Vortex Mixers.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
Vortex
Shipping Condition
Room Temperature
Product Line
Ambion™
Type
Tools and Accessories

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text