Single Place Magnetic Stand (for one 1.5 mL tube)
Single Place Magnetic Stand (for one 1.5 mL tube)

Single Place Magnetic Stand (for one 1.5 mL tube)

The Ambion® Single Tube Magnetic Stand accommodates one 1.5 mL microfuge tube. With this stand, as with any magnetic stand,Read more
Have Questions?
Catalog NumberQuantity
AM100261 Each
Catalog number AM10026
Price (USD)
134.65
Online Exclusive
140.00
Save 5.35 (4%)
Each
Add to cart
Quantity:
1 Each
Price (USD)
134.65
Online Exclusive
140.00
Save 5.35 (4%)
Each
Add to cart
Ask our AI about this Product
The Ambion® Single Tube Magnetic Stand accommodates one 1.5 mL microfuge tube. With this stand, as with any magnetic stand, centrifugation is replaced by the attraction between capture beads in a solution and the magnet in the stand, resulting in the beads collecting on one side of the microfuge tube. Thus, the captured species (e.g., mRNA) is quickly and efficiently separated from the solution's other components.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Magnetic Stand
Quantity1 Each
Shipping ConditionRoom Temperature
Product LineAmbion™
TypeMagnetic Stand
Unit SizeEach
Contents & Storage
Store at room temperature.
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Figures

Customers who viewed this item also viewed



Frequently asked questions (FAQs)

Ambion magnetic stands (AM10050 & AM10026) are not designed for and do not work well for 0.2 mL individual PCR tubes or strips.

Share catalog number, name or link

1x1 image pixel for data collection