Informational:Inquire about OEM or Commercial Supply version of this product here.

Magnetic Stand-96

Catalog number: AM10027
Have Questions?

Magnetic Stand-96

Catalog number: AM10027
Have Questions?
Catalog Number
AM10027
Unit Size
1 each
Price (USD)
Availability
-
Quantity
Catalog NumberUnit SizePrice (USD)AvailabilityQuantity
AM100271 each
-
Product Overview
Figures
Videos
Recommendations
Recommendations
Documents
FAQ
Citations & References
Additional Information
Recommendations
The Magnetic Stand-96 is designed for paramagnetic bead precipitation from standard 96-well, U-bottom microplates and 0.2 mL PCR plates with no additional accessories. The stand has a white polyoxymethylene (POM) base in standard SBS 96-well microplate footprint. The use of 24 extremely strong permanent magnets guarantees easy and fast magnetic separation in as little as 30 seconds. It sits flat on a lab bench as well as in a microplate adapter on robotic workstations.

Using the Magnetic Stand-96
The Magnetic Stand-96 is ideal for use with MagMAX™-96 magnetic bead-based RNA isolation kits, MagMAX™ for Stabilized Blood Tubes RNA Isolation Kits, MagMAX™ FFPE Nucleic Acid Isolation Kits, and the MessageAmp™ Premier RNA Amplification Kit.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment)
Magnetic Stand
Shipping Condition
Room Temperature
Product Line
Ambion™
Quantity
1 Each
Type
Magnetic Stand

Contents & Storage

Store at room temperature.

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text